Skip to Content
Merck
All Photos(1)

Key Documents

1.10398

Millipore

GranuCult® FRASER broth (base)

pkg of 500 g, according to ISO 11290, APHA, selective for Listeria spp.

Synonym(s):

Listeria Selective Enrichment Broth, Full FRASER broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

APHA
according to ISO 11290

Quality Level

sterility

non-sterile

form

granular

packaging

pkg of 500 g

manufacturer/tradename

GranuCult® prime

storage condition

protect from light (both UV and Sunlight)

technique(s)

microbiological culture: suitable

pH

7.0-7.4 (25 °C, 57.4 g/L in H2O)

solubility

57.4 g/L

application(s)

food and beverages
microbiology
pathogen testing

storage temp.

15-25°C

suitability

selective for Listeria spp.

General description

GranuCult® FRASER broth (base) has high nutrient content and a large buffering capacity that creates optimum growth conditions for Listeria spp. Lithium chloride, nalidixic acid, and acriflavine hydrochloride inhibit the growth of accompanying bacteria. The addition of aesculin and ammonium iron (III) citrate facilitates the detection of the β-D-glucosidase activity by forming a black complex of aesculin with iron (III) ions. A black coloration can develop during incubation, but this reaction is not exclusive to Listeria spp. and cultures that show no blackening cannot be assumed to be free of Listeria spp. Hence, every primary and secondary enrichment in Fraser broth must be sub-cultured on selective plating media. From GranuCult® FRASER Broth (Base), the primary selective enrichment Half Fraser broth, as well as the secondary enrichment Fraser broth can be prepared by addition of Ammonium iron (III) citrate supplement (1.00092) and Selective Supplement (1.00093).

Application

GranuCult® FRASER broth (base) is suitable for the primary and secondary selective enrichment of Listeria spp. from food and animal feed as well as from environmental samples and other materials.

Linkage

Replaces: 1.10398

Analysis Note

Appearance (clearness): clear to almost clear
Appearance (color): yellowish-brown
pH-value (25 °C): 7.0 - 7.4
Growth promotion test in accordance with the current version of DIN EN ISO 11133.
Mixed inoculum of Listeria, E.coli and E.faecalis
Additionaly monoculture of E.coli and E.faecalis
Confirmation on ALOA®-Medium resp. Tryptic soy agar
Inoculum on reference medium (Listeria monocytogenes ATCC 13932 (WDCM 00021)): ≤ 100
Inoculum on reference medium (Listeria monocytogenes ATCC 35152 (WDCM 00109)): ≤ 100
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)): ≥ 1000
Inoculum on reference medium (Escherichia coli ATCC 8739 (WDCM 00012)): ≥ 1000
Inoculum on reference medium (Enterococcus faecalis ATCC 19433 (WDCM 00009)): ≥ 1000
Inoculum on reference medium (Enterococcus faecalis ATCC 29212 (WDCM 00087)): ≥ 1000
Blacking (Listeria monocytogenes ATCC 13932 (WDCM 00021)): +
Blacking (Listeria monocytogenes ATCC 35152 (WDCM 00109)): +
Blacking (Escherichia coli ATCC 25922 (WDCM 00013)): -
Blacking (Escherichia coli ATCC 8739 (WDCM 00012)): -
Blacking (Enterococcus faecalis ATCC 19433 (WDCM 00009)): -
Blacking (Enterococcus faecalis ATCC 29212 (WDCM 00087)): -
Growth on ALOA medium (Listeria monocytogenes ATCC 13932 (WDCM 00021)): >10 colonies
Growth on ALOA medium(Listeria monocytogenes ATCC 35152 (WDCM 00109)): >10 colonies
Colonies (Listeria monocytogenes ATCC 13932 (WDCM 00021)): blue green colonies with opaque halo
Colonies(Listeria monocytogenes ATCC 35152 (WDCM 00109)): blue green colonies with opaque halo
Growth on Tryptic soy agar (Escherichia coli ATCC 25922 (WDCM 00013)): total inhibition
Growth on Tryptic soy agar (Escherichia coli ATCC 8739 (WDCM 00012)): total inhibition
Growth on Tryptic soy agar (Enterococcus faecalis ATCC 29212 (WDCM 00087)): <100 colonien
Growth on Tryptic soy agar (Enterococcus faecalis ATCC 19433 (WDCM 00009)): <100 colonies
Incubation:
Half-Fraser Broth: 24 ± 2 hours at 30 ± 1°C
Fraser Broth: 48 ± 2 hours at 37 ± 1°C
Confirmation on ALOA medium: 44 ± 4 hours at 37 ±1°C
Confirmation on Tryptic soy agar: 24 ± 3 hours at 37 ± 1°C

Legal Information

ALOA is a registered trademark of Biolife Italiana S.r.l.
GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered®culture media, depending on your needs. The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

Microbial culture media is available in both powdered and granulated forms. This article compares the characteristics of each culture media format with regards to safety, handling and convenience.

See All

Protocols

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Listeria is a public health concern, affecting elderly, pregnant, newborns, and immunocompromised individuals.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service