Skip to Content
Merck

1.10338

Millipore

Fractogel® EMD Chelate (M)

purified by immobilized metal affinity chromatography (IMAC)

Synonym(s):

Fractogel® EMD Chelate (M)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
Pricing and availability is not currently available.

ligand

iminodiacetic acid

Quality Level

description

for metal affinity chromatography (IMAC), suspension in 20% ethanol and 150 mM NaCl (40-90 µm)

sterility

sterile (Caustic Stable)

form

resin

purified by

immobilized metal affinity chromatography (IMAC)

manufacturer/tradename

Fractogel®

parameter

8 bar max. pressure
~240 cm/hr flow rate

matrix active group

methacrylate

wide

40-90 μm

capacity

~60 mg binding capacity( lysozyme/ml of resin)

transition temp

flash point 35 °C (calculated)

application(s)

recombinant protein

separation technique

affinity

storage temp.

2-30°C

General description

Fractogel® EMD Chelate combines our tentacle structure with iminodiacetic acid as the metal chelating ligand

Features and Benefits

Fractogel® EMD Chelate (M) enables users to:

  • Efficiently purify histidine-tagged recombinant proteins
  • Coordinate metal ions to non-bonding electron pairs of the amino acid side chains and separate peptides
  • Ensure timely and successful startup and operation
  • Minimize hydrophobic interactions due to the tentacles technology, resulting in superior selectivity and recovery
  • Handle multiple CIP and packing cycles with the chemically and physically robust bead, leading to a longer resin lifetime

Packaging

  • 1.10338.0100: Fractogel® EMD Chelate (M) Resin 100ml
  • 1.10338.0010: Fractogel® EMD Chelate (M) Resin 10 ml
  • 1.10338.0500: Fractogel® EMD Chelate (M) Resin 500 ml
  • 1.10338.5000: Fractogel® EMD Chelate (M) Resin 5 L

Plastic bottle

Analysis Note

Appearance: Milky tubid suspension, free from impurities (foreigen particles)
Microscopic evaluation: Uniform spherical particles, no agglomerates, no fines
Cerium: ≤ 10 µg/g
Copper-(II)-binding capacity: 60 - 100 µmol/ml
Extractable matter (water): ≤ 0.05 %
Pressure drop(column: ID=1.6 cm, L=10 cm at 5 ml/min): ≤ 1.0 bar
Particle size (d10): 37 - 45 µm
Particle size (d50): 48 - 60 µm
Particle size (d90): 63 - 77 µm
Colony forming units (TAMC + TYMC): ≤ 100 CFU/ml
Endotoxins: ≤ 1.00 EU/ml
Functional test: passes test
Functional test: Separation of chymotrypsinogen A and myoglobin

Legal Information

FRACTOGEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F

Flash Point(C)

35 °C


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service