Skip to Content
Merck
All Photos(1)

Key Documents

1.09494

Supelco

ICP multi-element standard solution IX

(9 elements in dilute nitric acid) 100 mg/l: As, Be, Cd, Cr(VI), Hg, Ni, Pb, Se, Tl Certipur®

Synonym(s):

Multi-Element Calibration Solution for ICP/AAS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

reference material

Quality Level

product line

Certipur®

technique(s)

ICP: suitable

pH

0.3 (20 °C in H2O)

bp

100 °C/1013 hPa

density

1.050 g/cm3 at 20 °C

application(s)

industrial qc
pharmaceutical

format

multi-component solution

storage temp.

15-25°C

Application

  • ICP multi-element calibration standard: Though specific PubMed references are not available, ICP multi-element standard solution IX is critical in trace metal analysis, providing a reliable calibration standard for ICP-MS, ensuring accuracy in measuring elemental impurities in pharmaceutical products.
  • Trace metal analysis standard for pharmaceutical quality control: This solution is essential in the pharmaceutical industry for ensuring compliance with regulatory requirements on elemental impurities, serving as a key component in quality control laboratories.
  • Elemental analysis reagent in biochemical research: ICP multi-element standard solution IX is invaluable for researchers in biochemistry and biotechnology, facilitating the precise quantification of trace elements in various biological samples and supporting studies on their physiological roles and impacts.

Analysis Note

As (arsenic): 100 mg/l
Be (beryllium): 100 mg/l
Pb (lead): 100 mg/l
Cd (cadmium): 100 mg/l
Cr (chromium): 100 mg/l
Ni (Nickel): 100 mg/l
Hg (Mercury): 100 mg/l
Se (Selenium): 100 mg/l
Tl (Thallium): 100 mg/l

Other Notes

For a complete product listing of our ICP and AAS range, including technical information and example certificates, please visit ICP & AAS standards website.

Detailed information on our complete workflow solutions for ICP-AAS can be found in our Brochure: High-Purity Chemicals for Inorganic Trace Analysis.

In our effort to be more sustainable, we′re going digital with COAs-download them directly from our website.

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service