1.07062
Diethyl ether
ethanol stabilized, EMPARTA® ACS reagent
Synonym(s):
Ethyl ether, Ether, Et2O, Ethoxyethane, Ether, Ethyl ether
About This Item
Recommended Products
Product Name
Diethyl ether, for analysis, Ethanol stabilized EMPARTA® ACS
grade
ACS reagent
Quality Level
vapor density
2.6 (vs air)
vapor pressure
587 hPa ( 20 °C)
product line
EMPARTA®
form
liquid
autoignition temp.
320 °F
potency
1215 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)
expl. lim.
36.5 %
dilution
(for routine analytical applications)
refractive index
n20/D 1.3530 (lit.)
bp
34.6 °C (lit.)
mp
−116 °C (lit.)
transition temp
flash point -40 °C
density
0.706 g/mL at 25 °C (lit.)
storage temp.
15-25°C
SMILES string
CCOCC
InChI
1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3
InChI key
RTZKZFJDLAIYFH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
-40.0 °F - closed cup
Flash Point(C)
-40 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service