Skip to Content
Merck
All Photos(4)

Key Documents

1.06504

Millipore

Sodium deoxycholate

Bile salt and ionic detergent. Supplement. For the isolation and classification of gram-negative bacteria

Synonym(s):

Sodium deoxycholate, Deoxycholic acid sodium salt, Desoxycholic acid sodium salt 

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H39NaO4
CAS Number:
Molecular Weight:
414.55
MDL number:
UNSPSC Code:
41106212
EC Index Number:
206-132-7
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

form

solid

potency

1370 mg/kg LD50, oral (Rat)

pH

7.5-9.0 (20 °C, 20 g/L in H2O)

mp

357-365 °C

transition temp

flash point >110 °C (own results)

solubility

330 g/L

bulk density

200 kg/m3

application(s)

microbiology

storage temp.

15-25°C

SMILES string

[Na+].[O-]C(=O)CC[C@H]([C@@H]1[C@@]2([C@H]([C@H]3[C@@H]([C@@]4([C@@H](C[C@@H](CC4)O)CC3)C)C[C@@H]2O)CC1)C)C

InChI

1S/C24H40O4.Na/c1-14(4-9-22(27)28)18-7-8-19-17-6-5-15-12-16(25)10-11-23(15,2)20(17)13-21(26)24(18,19)3;/h14-21,25-26H,4-13H2,1-3H3,(H,27,28);/q;+1/p-1/t14-,15-,16-,17+,18-,19+,20+,21+,23+,24-;/m1./s1

InChI key

FHHPUSMSKHSNKW-SMOYURAASA-M

General description

Sodium deoxycholate is a water-soluble, bile salt and an ionic detergent used as a selective inhibitory agent for bacteriological culture media. It interferes with the growth and incorporation of glucose into various cellular fractions and also inhibits flagellum formation in gram negative bacteria.

Application

Sodium deoxycholate is used in number of bacteriological media for the isolation and classification of gram-negative bacteria from food and the environment.

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 98.5 %
Identity (IR-spectrum): passes test
pH-value (2 %; water): 7.5 - 9.5
Spec. rotation (α 20/D; 2 %; water; calc. on dried substance):
Heavy metals (as Pb): ≤ 0.001 %
TLC-Test: passes test
Cholic acid (TLC): ≤ 2.0 %
Loss on drying (TG): ≤ 6.0 %
Suitability for microbiology (MacConkey-Agar): passes test
The substance meets the criteria described in the monograph1483 Ph.Eur. 6th Ed. and any subsequently revised version.
For details, please request our BSE-certificate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service