Skip to Content
Merck
All Photos(2)

Key Documents

1.02827

Supelco

Cyclohexane

for liquid chromatography LiChrosolv®

Synonym(s):

Hexahydrobenzene, Hexamethylene, Naphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-806-2
NACRES:
NA.03
Assay:
≥99.9% (GC)
technique(s):
HPLC: suitable
bp:
80.7 °C (lit.)
vapor pressure:
168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

2.9 (vs air)

Quality Level

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

description

isocratic

product line

LiChrosolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

500 °F

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

9 %

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.01% Water

evapn. residue

≤2.0 mg/L

transmittance

230 nm, ≥75%
240 nm, ≥90%
260 nm, ≥99%

refractive index

n20/D 1.426 (lit.)

kinematic viscosity

1.26 cSt(20 °C)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

transition temp

flash point -18 °C

density

0.779 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.

Application


  • Phytochemical Profiling and Biological Potential of Prunus dulcis Shell Extracts.: Cyclohexane is utilized here for the extraction and analysis of bioactive compounds in almond shells, illustrating its application in natural product chemistry and analytical methodologies. The study underscores cyclohexane′s effectiveness in extracting valuable phytochemicals for potential therapeutic and commercial use (Ben Khadher T et al., 2023).

  • Determining the Antibacterial Effect of Mentha Longifolia Essential Oil on Cariogenic Bacteria and Its Compounds: an in vitro Study.: In this study, cyclohexane is employed as a solvent to isolate essential oils for antibacterial testing, highlighting its critical role in enhancing the efficacy of natural antimicrobial agents. The research contributes to the development of cyclohexane-based methods for studying and applying essential oils in dental health (Shazdehahmadi F et al., 2023).


Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.9 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.01 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 230 nm): ≥ 75 %
Transmission (at 240 nm): ≥ 90 %
Transmission (from 260 nm): ≥ 99 %
Filtered by 0.2 µm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service