Skip to Content
Merck
All Photos(1)

Key Documents

1.01878

Supelco

Bismuth(III) nitrate

alkaline for analysis EMSURE® Reag. Ph Eur

Synonym(s):

Bismuth(III) nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bi5O(OH)9(NO3)4
CAS Number:
Molecular Weight:
1461.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
215-136-8
NACRES:
NA.21
form:
solid

Agency

reag. Ph. Eur.

Quality Level

product line

EMSURE®

Assay

≥71.0% Bi basis (complexometric)

form

solid

impurities

≤0.5% Substances not precipitated by hydrogen sulfide (as sulfate)

pH

3.2 (20 °C, 50 g/L in H2O, slurry)

density

4.93 g/cm3 at 20 °C

bulk density

600 kg/m3

anion traces

chloride (Cl-): ≤0.005%

cation traces

Ag: ≤0.001%
As: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.002%
Fe: ≤0.002%
NH4+: ≤0.01%
Pb: ≤0.005%
Zn: ≤0.001%

storage temp.

2-30°C

SMILES string

[Bi](O[N+](=O)[O-])(O[N+](=O)[O-])O[N+](=O)[O-]

InChI

1S/Bi.3NO3/c;3*2-1(3)4/q+3;3*-1

InChI key

PPNKDDZCLDMRHS-UHFFFAOYSA-N

Related Categories

Application


  • In-house validation method for quantification of formaldehyde in silage and feedingstuffs with the use HPLC-DAD technique.: Demonstrating the versatility of 2,4-Dinitrophenylhydrazine phosphoric acid solutions in food safety, this paper describes a validated method for detecting formaldehyde residues in animal feed, ensuring compliance with health and safety regulations (Patyra et al., 2020).

  • Development of a measurement method to determine the ceiling exposure concentration of ortho-phthalaldehyde handling workers.: This research uses derivatization agents including 2,4-Dinitrophenylhydrazine phosphoric acid solution to measure harmful substances in occupational settings, contributing to enhanced worker protection and industrial hygiene (Yamamoto et al., 2020).

  • Reductive amination of aldehyde 2,4-dinitorophenylhydrazones using 2-picoline borane and high-performance liquid chromatographic analysis.: Focusing on the chemical synthesis processes, this study employs 2,4-Dinitrophenylhydrazine phosphoric acid solutions to analyze reaction mechanisms in pharmaceutical and biotechnological research, fostering advancements in drug development and synthetic chemistry (Uchiyama et al., 2009).


Analysis Note

Assay (complexometric, Bi): ≥ 71.0 %
Chloride (Cl): ≤ 0.005 %
Ag (Silver): ≤ 0.001 %
As (Arsenic): ≤ 0.0005 %
Ca (Calcium): ≤ 0.005 %
Cu (Copper): ≤ 0.002 %
Fe (Iron): ≤ 0.002 %
NH₄ (Ammonium): ≤ 0.01 %
Pb (Lead): ≤ 0.005 %
Zn (Zinc): ≤ 0.001 %
Substances not precipitated by hydrogen sulfide (as sulfate): ≤ 0.5 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service