Skip to Content
Merck
All Photos(2)

Key Documents

1.01216

SAFC

Ammonium sulfate

extra pure

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
MDL number:
UNSPSC Code:
12352301
EC Index Number:
231-984-1
Pricing and availability is not currently available.

vapor pressure

<1 Pa ( 25 °C)

Quality Level

Assay

≥99.0% (acidimetric)

form

solid

potency

2840 mg/kg LD50, oral (Rat)

pH

5 (100 g/L in H2O)

mp

>280 °C (dec.) (lit.)

solubility

754-764 g/L

density

1.77 g/mL at 25 °C (lit.)

bulk density

850 kg/m3

application(s)

pharma/biopharma processes

storage temp.

2-30°C

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Ammonium sulfate is used for large and laboratory scale protein purification and fractionation that can be applied to separate proteins by altering their solubility in the presence of a high salt concentration.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service