1.00570
di-Phosphorus pentoxide
for analysis ACS,ISO,Reag. Ph Eur
Synonym(s):
di-Phosphorus pentoxide, Phosphoric anhydride
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Agency
reag. ISO
reag. Ph. Eur.
vapor pressure
<0.1 hPa ( 20 °C)
Assay
≥98% (acidimetric)
form
solid
impurities
≤0.01% reductants (as P2O5)
≤20 ppm nitrogen (N)
≤50 ppm insolubles
pH
1 (20 °C, 5 g/L in H2O)
mp
420 °C (closed capillary tube)
density
2.28 g/cm3 at 20-25 °C
bulk density
700 kg/m3
anion traces
chloride (Cl-): ≤10 ppm
nitrate (NO3-): ≤5 ppm
cation traces
As: ≤200 ppm
Ca: ≤50 ppm
Cd: ≤5 ppm
Co: ≤5 ppm
Cu: ≤5 ppm
Fe: ≤10 ppm
K: ≤20 ppm
Mg: ≤10 ppm
Mn: ≤5 ppm
NH4+: ≤10 ppm
Na: ≤50 ppm
Ni: ≤5 ppm
Pb: ≤5 ppm
Sb: ≤10 ppm
Zn: ≤5 ppm
heavy metals (as Pb): ≤50 ppm
storage temp.
no temp limit
SMILES string
[P]21(=O)O[P]3(=O)O[P](=O)(O2)O[P](=O)(O1)O3
InChI
1S/O10P4/c1-11-5-12(2)8-13(3,6-11)10-14(4,7-11)9-12
InChI key
DLYUQMMRRRQYAE-UHFFFAOYSA-N
Application
- Synthesis of new azo compounds based on N-(4-hydroxypheneyl)maleimide and N-(4-methylpheneyl)maleimide.: This study presents the synthesis of novel azo compounds utilizing di-Phosphorus pentoxide, demonstrating its utility in facilitating complex reactions in organic synthesis, particularly in forming N-substituted maleimide derivatives crucial in analytical chemistry for detecting molecular interactions (Mohammed IA et al., 2010).
Analysis Note
Identity: passes test
Insoluble matter: ≤ 50 ppm
Heavy metals (as Pb): ≤ 50 ppm
Total nitrogen (N): ≤ 20 ppm
Chloride (Cl): ≤ 10 ppm
Nitrate (NO₃): ≤ 5 ppm
As (Arsenic): ≤ 200 ppm
Ca (Calcium): ≤ 50 ppm
Cd (Cadmium): ≤ 5 ppm
Co (Cobalt): ≤ 5 ppm
Cu (Copper): ≤ 5 ppm
Fe (Iron): ≤ 10 ppm
K (Potassium): ≤ 20 ppm
Mg (Magnesium): ≤ 10 ppm
Mn (Manganese): ≤ 5 ppm
NH₄ (Ammonium): ≤ 10 ppm
Na (Sodium): ≤ 50 ppm
Ni (Nickel): ≤ 5 ppm
Pb (Lead): ≤ 5 ppm
Sb (Antimony): ≤ 10 ppm
Zn (Zinc): ≤ 5 ppm
Reducing matter (P₂O₃): ≤ 0.01 %
Corresponds to ACS,ISO,Reag. Ph Eur
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1A
Supplementary Hazards
Storage Class Code
8B - Non-combustible, corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service