Skip to Content
Merck
All Photos(2)

Key Documents

1.00383

SAFC

DL-Malic acid

EMPROVE® ESSENTIAL, Ph. Eur., NF, FCC, E 296

Pharma Manufacturing

Synonym(s):

DL-Malic acid, DL-Hydroxysuccinic acid, Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O5
CAS Number:
Molecular Weight:
134.09
MDL number:
UNSPSC Code:
12352106
E Number:
E296
EC Index Number:
230-022-8

Agency

NF
Ph. Eur.

Quality Level

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

solid

autoignition temp.

349 °C

potency

>3200 mg/kg LD50, oral (Rat)

technique(s)

API processing | cocrystal formation: suitable
API processing | salt formation: suitable

pH

2.3 (20 °C, 10 g/L in H2O)

mp

128-132 °C

transition temp

flash point 203 °C

solubility

558 g/L

density

1.60 g/cm3 at 20 °C

application(s)

pharmaceutical
solid formulation

storage temp.

15-25°C

SMILES string

[O-]C(=O)C(O)CC(=O)[O-].[H+].[H+]

InChI

1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)

InChI key

BJEPYKJPYRNKOW-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our EMPROVE® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Malic acid is typically applied as acidulant and pH adjuster as well as taste modifier in pharmaceutical formulations and food. In addition, it has antioxidant properties.
DL-Malic acid Emprove® Essential is mainly used in highly regulated pharmaceutical applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

397.4 °F

Flash Point(C)

203 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service