Skip to Content
Merck
All Photos(2)

Key Documents

1.00313

Supelco

Hydrochloric acid

32%, EMPLURA®

Synonym(s):

Hydrochloric acid solution, Hydrochloric acid 32%, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Hydrochloric acid solution, 32% EMPLURA®

vapor pressure

21.3 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥32% (alkalimetric)

form

liquid

impurities

≤0.0001% As (Arsenic)
≤0.0001% Free chlorine (Cl)
≤0.0001% Heavy metals (as Pb)
≤0.0002% Fe (Iron)
≤0.0005% Sulfate (SO4)
≤0.001% Sulfite (SO?)

pH

<1 (20 °C in H2O)

mp

-50 °C

density

1.16 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Research on the Synthesis of Zinc-Ammonium Phosphate Using Galvanic Waste Sludge as a Source of Zinc: This study explores a novel approach using hydrochloric acid solution in the synthesis process aimed at valorizing waste materials. The article demonstrates a chemical strategy that enhances resource recovery, significantly contributing to waste minimization in industrial practices (Morgovan et al., 2024).

  • Contactless conductivity sensor employing moist paper as absorbent for in-situ detection of generated carbon dioxide gas: The study integrates hydrochloric acid solutions in the development of a novel, cost-effective contactless conductivity sensor. This sensor uses a simple paper-based system for the in-situ monitoring of gases, providing a significant tool in environmental monitoring and analysis (Nacapricha et al., 2020).


Analysis Note

Assay (alkalimetric): ≥ 32 %
Identity: passes test
Bromide, Iodide (Br,I): passes test
Free chlorine (Cl): ≤ 0.0001 %
Sulfate (SO₄): ≤ 0.0005 %
Sulfite (SO₃): ≤ 0.001 %
Heavy metals (as Pb): ≤ 0.0001 %
As (Arsenic): ≤ 0.0001 %
Fe (Iron): ≤ 0.0002 %
Residual solvents (ICH Q3C): ...excluded by production process
Evaporation residue: ≤ 0,01
Expiry date : see product label

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service