09-278
Anti-20S proteasome subunit β5 Antibody
serum, Upstate®
Synonym(s):
LMP17, MB1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
manufacturer/tradename
Upstate®
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
human ... PSMB5(5693)
Related Categories
Specificity
Recognizes proteasome subunit β5
Immunogen
Peptide corresponding to residues 194-207 of human proteasome subunit β5 conjugated to KLH.
Application
Research Category
Protein Trafficking
Protein Trafficking
Research Sub Category
Ubiquitin & Ubiquitin Metabolism
Ubiquitin & Ubiquitin Metabolism
This Anti-20S proteasome subunit β5 Antibody is validated for use in IH, WB for the detection of 20S proteasome subunit β5.
Target description
22 kDa
Physical form
100 µL of purified antiserum preparation in PBS containing 0.09% sodium azide. Dilute to working strength with phosphate buffered saline pH 7.2-7.4.
antiserum
Storage and Stability
Stable for 1 year at -20°C from date of shipment for up to 12 months.
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
iScience, 12, 318-332 (2019-02-11)
Reduced poly(A)-binding protein nuclear 1 (PABPN1) levels cause aging-associated muscle wasting. PABPN1 is a multifunctional regulator of mRNA processing. To elucidate the molecular mechanisms causing PABPN1-mediated muscle wasting, we compared the transcriptome with the proteome in mouse muscles expressing short
Molecular biology reports, 24(1-2), 3-11 (1997-03-01)
The 26S proteasome is an eukaryotic ATP-dependent, dumbbell-shaped protease complex with a molecular mass of approximately 2000 kDa. It consists of a central 20S proteasome, functioning as a catalytic machine, and two large V-shaped terminal modules, having possible regulatory roles
PLoS genetics, 12(5), e1006031-e1006031 (2016-05-07)
Poly(A) Binding Protein Nuclear 1 (PABPN1) is a multifunctional regulator of mRNA processing, and its expression levels specifically decline in aging muscles. An expansion mutation in PABPN1 is the genetic cause of oculopharyngeal muscle dystrophy (OPMD), a late onset and
null
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service