Skip to Content
Merck
All Photos(4)

Key Documents

07-732

Sigma-Aldrich

Anti-HDAC6 Antibody, CT

Upstate®, from rabbit

Synonym(s):

Anti-CPBHM, Anti-HD6, Anti-JM21, Anti-PPP1R90

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HDAC6(10013)

Specificity

HDAC6

Immunogen

6His-tagged fusion protein corresponding to residues 1031-1215 of human histone deacetylase 6 (HDAC6).

Application

Anti-HDAC6 Antibody, CT is a Rabbit Polyclonal Antibody for detection of HDAC6 also known as histone deacetylase 6 & has been tested in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by immunoblot.

Target description

134kDa

Linkage

Replaces: MABE546

Physical form

Format: Purified
Protein A purified

Storage and Stability

2 years at -20°C from date of shipment

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homer2 deletion alters dendritic spine morphology but not alcohol-associated adaptations in GluN2B-containing N-methyl-D-aspartate receptors in the nucleus accumbens.
McGuier, NS; Padula, AE; Mulholland, PJ; Chandler, LJ
Frontiers in Pharmacology null
Withdrawal from chronic intermittent alcohol exposure increases dendritic spine density in the lateral orbitofrontal cortex of mice.
McGuier, NS; Padula, AE; Lopez, MF; Woodward, JJ; Mulholland, PJ
Alcohol null
Epigenetic status determines germ cell meiotic commitment in embryonic and postnatal mammalian gonads.
Wang, N; Tilly, JL
Cell Cycle null
Upregulation of miR-22 promotes osteogenic differentiation and inhibits adipogenic differentiation of human adipose tissue-derived mesenchymal stem cells by repressing HDAC6 protein expression.
Huang, S; Wang, S; Bian, C; Yang, Z; Zhou, H; Zeng, Y; Li, H; Han, Q; Zhao, RC
Stem Cells and Development null
Hui Su Jeong et al.
Molecules and cells, 45(4), 231-242 (2022-04-01)
The neuromuscular junction (NMJ), which is a synapse for signal transmission from motor neurons to muscle cells, has emerged as an important region because of its association with several peripheral neuropathies. In particular, mutations in GARS that affect the formation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service