Skip to Content
Merck
All Photos(5)

Documents

07-478

Sigma-Aldrich

Anti-SNF2β/BRG1 Antibody

serum, Upstate®

Synonym(s):

Probable global transcription activator SNF2L4, ATP-dependent helicase SMARCA4, SNF2-beta, BRG-1, Mitotic growth and transcription activator, Brahma protein homolog 1, SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily A m

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... SMARCA4(6597)

General description

The SNF2 family of proteins are ATP dependant chromatin remodeling enzymes. The gene encoding this protein is also refered to as SMARCA4.
The SWI-SNF complex is involved in the activation of transcription via the remodeling of nucleosome structure in an ATP-dependent manner. Brm (also designated SNF2α) and Brg-1 (also designated SNF2β) are the ATPase subunits of the mammalian SWI-SNF complex. Brm, Brg-1, Ini1 (integrase interactor 1, also designated SNF5), BAF155 (also designated SRG3) and BAF170 are thought to comprise the functional core of the SWI-SNF complex. Addition of Ini1, BAF155 and BAF170 to Brg-1 appears to increase remodeling activity. Other complex subunits are thought to play regulatory roles. hSNF2L and hSNF2H both appear to be homologs of Drosophila ISWI, a Brm related ATPase that is present in chromatin remodeling complexes other than SWI/SNF, including the NURF (nucleosome remodeling factor).

Specificity

This antibody recognizes the Near N-Terminus of SNF2B

Immunogen

GST-tagged recombinant protein corresponding to the Near N-Terminus of human SNF2β.

Application

Immunocytochemistry Analysis: A 1:250 dilution from a representative lot detected BRG1 in HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair

Transcription Factors
Use Anti-SNF2β/BRG1 Antibody (rabbit polyclonal antibody) validated in IP, WB to detect SNF2β/BRG1 also known as Probable global transcription activator SNF2L4, ATP-dependent helicase SMARCA4, SNF2-beta, BRG-1.

Quality

Evaluated by Western Blotting in HELA NE cell lysate.
Western Blotting Analysis: A 1:4,000 dilution of this antibody detected SNF2B/BRG1 in 10 µg of HELA NE cell lysate.

Target description

200 kDa

Physical form

Rabbit polyclonal in buffer containing with 0.05% sodium azide with 30% glycerol.
Unpurified

Storage and Stability

Maintain for 2 years at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-309, Hela cell nuclear extract. Add an equal volume of Laemmli reducing sample buffer to 10 μL of extract and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced extract per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Germline and somatic SMARCA4 mutations characterize small cell carcinoma of the ovary, hypercalcemic type.
Witkowski, Leora, et al.
Nature Genetics, 46, 438-443 (2014)
BRG1 increases transcription of proinflammatory genes in renal ischemia.
Naito, M; Zager, RA; Bomsztyk, K
Journal of the American Society of Nephrology null
Petar Jelinic et al.
Nature genetics, 46(5), 424-426 (2014-03-25)
Small cell carcinoma of the ovary, hypercalcemic type (SCCOHT) is a rare, highly aggressive form of ovarian cancer primarily diagnosed in young women. We identified inactivating biallelic SMARCA4 mutations in 100% of the 12 SCCOHT tumors examined. Protein studies confirmed
Combinatorial probabilistic chromatin interactions produce transcriptional heterogeneity.
Voss, TC; Schiltz, RL; Sung, MH; Johnson, TA; John, S; Hager, GL
Journal of Cell Science null
Joel Vincent Fair et al.
BMC developmental biology, 16(1), 27-27 (2016-08-04)
The Hedgehog (HH) signalling pathway regulates cardiomyogenesis in vivo and in differentiating P19 embryonal carcinoma (EC) cells, a mouse embryonic stem (mES) cell model. To further assess the transcriptional role of HH signalling during cardiomyogenesis in stem cells, we studied

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service