Skip to Content
Merck
All Photos(1)

Documents

07-412

Sigma-Aldrich

Anti-dimethyl-Arginine Antibody, symmetric (SYM10)

serum, Upstate®

Synonym(s):

Anti-SYM10, Dimethyl-Arginine Detection Ab, Symmetric Dimethyl-Arginine Ab

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

target post-translational modification

dimethylation (Arg)

Related Categories

General description

Arginine methylation is a post-translational modification that results in the formation of asymmetrical and symmetrical dimethylated arginines (a- and sDMA).
Does not recognize non-methylated peptides, peptides containing asymmetrically dimethylated arginines, or a peptide containing a single symmetric dimethylarginine.

Specificity

Recognizes proteins that contain multiple symmetrically dimethylated arginines, including p80-Coilin and the Sm protein B, B′ and D.

Immunogen

peptide containing four symmetrical dimethyl-arginine-glycine repeats

Application

Anti-dimethyl-Arginine Antibody, symmetric (SYM10) is a high quality Rabbit Polyclonal Antibody for the detection of dimethyl-Arginine & has been validated in ICC, IP & WB.
Research Category
Signaling
Research Sub Category
Chromatin Biology

Quality

routinely evaluated by immunoblot in RIPA lysates from Jurkat cells

Target description

varies

Physical form

Whole rabbit antiserum containing 0.05% sodium azide and 30% glycerol. Liquid at -20°C.

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-303, Jurkat cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lindsay M Webb et al.
Frontiers in immunology, 10, 524-524 (2019-04-04)
Multiple sclerosis is an autoimmune disease of the central nervous system (CNS) mediated by CD4+ T cells and modeled via experimental autoimmune encephalomyelitis (EAE). Inhibition of PRMT5, the major Type II arginine methyltransferase, suppresses pathogenic T cell responses and EAE.
Arginine methylation mediated by the Arabidopsis homolog of PRMT5 is essential for proper pre-mRNA splicing.
Deng, X; Gu, L; Liu, C; Lu, T; Lu, F; Lu, Z; Cui, P; Pei, Y; Wang, B; Hu, S; Cao, X
Proceedings of the National Academy of Sciences of the USA null
Madhumitha Rengasamy et al.
Nucleic acids research, 45(19), 11106-11120 (2017-10-05)
We observed overexpression and increased intra-nuclear accumulation of the PRMT5/WDR77 in breast cancer cell lines relative to immortalized breast epithelial cells. Utilizing mass spectrometry and biochemistry approaches we identified the Zn-finger protein ZNF326, as a novel interaction partner and substrate
Hironari Tamiya et al.
The Journal of clinical investigation, 128(1), 517-530 (2017-12-12)
SHARPIN, an adaptor for the linear ubiquitin chain assembly complex (LUBAC), plays important roles in NF-κB signaling and inflammation. Here, we have demonstrated a LUBAC-independent role for SHARPIN in regulating melanoma growth. We observed that SHARPIN interacted with PRMT5, a
Joris Paris et al.
Retrovirology, 15(1), 48-48 (2018-07-13)
Nuclear localization of Gag is a property shared by many retroviruses and retrotransposons. The importance of this stage for retroviral replication is still unknown, but studies on the Rous Sarcoma virus indicate that Gag might select the viral RNA genome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service