Skip to Content
Merck
All Photos(1)

Key Documents

07-1491

Sigma-Aldrich

Anti-Calcineurin pan A Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

CAM-PRP catalytic subunit, Calmodulin-dependent calcineurin A subunit alpha isoform, calcineurin A alpha, protein phosphatase 3 (formerly 2B), catalytic subunit,

alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit,

alp

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

100
200

conjugate

conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

purified by

affinity chromatography

species reactivity

human, rat, mouse

species reactivity (predicted by homology)

mammals (based on 100% sequence homology)

technique(s)

western blot: suitable

immunogen sequence

KLH-conjugated synthetic peptide corresponding to Calcineurin.

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

General description

Calcineurin is a serine/threonine protein phosphatase involved in the mediation of signal tranduction in a wide variety of cellular responses. Calcineurin has been linked to Pmk1 MAP kinase and PI pathways, as well as to members of the small GTPase Rab/Ypt family and Type II myosin. Furthermore, calcineurin participates in the transduction of extracellular signals to the nucleus by dephosphorylation of members of the NFAT family of transcription factors. These linkages demonstrate calcineurin′s multifunctional capabilities in the regulation of calcium signaling.

Specificity

This antibody recognizes both the Aα and Aβ, the major catalytic subunit gene isoforms of calcineurin in most tissues; may also react with the testis form, Aγ. This antibody has very high sensitivity and specificity, which allow detection of calcineurin in tissues where expression is low, such as lymphoid tissue. Reacts with calcineurin of all mammalian species.

Immunogen

KLH-conjugated synthetic peptide corresponding to Calcineurin.

Application

Detect Calcineurin pan A using this Anti-Calcineurin pan A Antibody validated for use in WB.
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling

Quality

Evaluated by Western Blot in Mouse brain lysate.
Western Blot Analysis: 1:1,000 dilution of this antibody detected Calcineurin on 10 µg of mouse brain lysate.

Target description

~58 kDa

Linkage

Replaces: AB1695

Physical form

Affinity purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH7.4) 150 mM NaCl with 40% gylerol. Contains no preservative.

Storage and Stability

Store at -20°C for up to 12 months in undiluted aliquots; at this temperature the solution will
remain unfrozen.

Analysis Note

Control
Mouse brain lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kelly M Grimes et al.
Frontiers in physiology, 14, 1054169-1054169 (2023-02-04)
Introduction: The ribosomal protein L3-like (RPL3L) is a heart and skeletal muscle-specific ribosomal protein and paralogue of the more ubiquitously expressed RPL3 protein. Mutations in the human RPL3L gene are linked to childhood cardiomyopathy and age-related atrial fibrillation, yet the
Divergent effects of GLP-1 analogs exendin-4 and exendin-9 on the expression of myosin heavy chain isoforms in C2C12 myotubes.
Lina Wang,Feng Guo,Shi Wei,Ruqian Zhao
Peptides null
Bastian Bruns et al.
Nature cardiovascular research, 2(7), 645-655 (2023-07-01)
Takotsubo syndrome (TTS) is an acute heart failure syndrome that mimics the symptoms of acute myocardial infarction and is often preceded by emotional and/or physical stress. There is currently no treatment for TTS. Here we show that injection of 2.5 mg kg-1
Val Andrew Fajardo et al.
Brain and behavior, 6(6), e00470-e00470 (2016-05-03)
Phospholamban (PLN) and sarcolipin (SLN) are small inhibitory proteins that regulate the sarco(endo)plasmic reticulum Ca(2+)-ATPase (SERCA) pump. Previous work from our laboratory revealed that in the soleus and gluteus minimus muscles of mice overexpressing PLN (Pln (OE)), SERCA function was
Kiran Bala et al.
PLoS pathogens, 8(9), e1002927-e1002927 (2012-10-03)
Kaposi's Sarcoma (KS), caused by Kaposi's Sarcoma Herpesvirus (KSHV), is a highly vascularised angiogenic tumor of endothelial cells, characterized by latently KSHV-infected spindle cells and a pronounced inflammatory infiltrate. Several KSHV proteins, including LANA-1 (ORF73), vCyclin (ORF72), vGPCR (ORF74), vIL6

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service