Skip to Content
Merck
All Photos(1)

Key Documents

06-903

Sigma-Aldrich

Anti-PKA Antibody, NT

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
polyclonal
application:
WB
species reactivity:
hamster, bovine, rat, pig, mouse, human
technique(s):
western blot: suitable
citations:
9

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

hamster, bovine, rat, pig, mouse, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TYRO3(7301)

Specificity

PKA, and recombinant human alpha isoform of PKA

Immunogen

peptide (KKGSEQESVKEFLAK-C) based upon amino acids 8-22 of the human 41kDa alpha isoform of the catalytic subunit of PKA

Application

Anti-PKA Antibody, NT detects level of PKA & has been published & validated for use in WB.
Research Category
Signaling
Research Sub Category
Cytoskeletal Signaling

Quality

routinely evaluated by immunoblot on rat brain microsomal preparation or PC-12 RIPA cell lysates

Target description

41kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol added to 30%
Format: Purified
Protein A chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ho-Lin Chuang et al.
Environmental toxicology, 36(9), 1873-1879 (2021-06-06)
Epimedium, is used traditionally in Chinese medicine to treat infertility problems. In this study, we establish the cell model to elucidate the protective effect of epimedium against ES by analyzing the molecular relationship between mitochondrial dynamics and steroidogenesis and to
Zhaoqing Zheng et al.
Journal of neurophysiology, 101(5), 2539-2549 (2009-03-06)
The cyclic AMP-dependent protein kinase (PKA) signaling pathway has been shown to be important in mechanisms of synaptic plasticity, although its direct and downstream signaling effects are not well understood. Using an in vitro model of eyeblink classical conditioning, we
The Chinese herbal medicine Dai-Zong-Fang promotes browning of white adipocytes in vivo and in vitro by activating PKA pathway to ameliorate obesity.
Xu, et al.
Frontiers in Pharmacology, 14, 1176443-1176443 (2023)
Gregory Segala et al.
Nature communications, 10(1), 1833-1833 (2019-04-25)
In response to extracellular signals, many signalling proteins associated with the plasma membrane are sorted into endosomes. This involves endosomal fusion, which depends on the complexes HOPS and CORVET. Whether and how their subunits themselves modulate signal transduction is unknown.
Wanxu Guo et al.
Biology open, 10(5) (2020-10-14)
Podocytes are the major target in proteinuric kidney diseases such as diabetic nephropathy. The underlying molecular mechanisms by which high glucose (HG) results in podocyte damage remain unclear. This study investigated the regulatory role of Smad3, ezrin, and protein kinase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service