Skip to Content
Merck
All Photos(2)

Key Documents

06-600

Sigma-Aldrich

Anti-NR2B Antibody

Upstate®, from rabbit

Synonym(s):

N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 2B, N-methyl-D-aspartate receptor subunit 3, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat (based on 100% sequence homology)

species reactivity (predicted by homology)

monkey (based on 100% sequence homology), canine (based on 100% sequence homology), bovine (based on 100% sequence homology), human (based on 100% sequence homology), chimpanzee (based on 100% sequence homology)

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... Grin2B(537804)
chimpanzee ... Grin2B(473373)
dog ... Grin2B(494009)
human ... GRIN2B(2904)
mouse ... Grin2B(14812)
rat ... Grin2B(24410)
rhesus monkey ... Grin2B(698571)

General description

Glutamate receptor ionotropic, NMDA 2B (UniProt: Q01097; also known as GluN2B, Glutamate [NMDA] receptor subunit epsilon-2, N-methyl D-aspartate receptor subtype 2B, NMDAR2B, NR2B) is encoded by the Grin2b gene (Gene ID: 14812) in mouse. NMDA receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long term potentiation, an activity dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B or NR2B), NMDAR2C (GRIN2C), and NMDAR2D (GRIN2D). GRIN2B may be a candidate gene for the neurodegenerative disorder dentato-rubro-pallidoluysian atrophy (DRPLA). Properties of NMDAR include modulation by glycine, inhibition by Zn2+, voltage dependent Mg2+ blockade and high Ca2+ permeability. The involvement of NMDAR in the CNS has become a focus area for neurodegenerative diseases such as Alzheimer′s disease, epilepsy and ischemic neuronal cell death.

Specificity

This rabbit polyclonal antibody specifically detects NR2B. It targets an epitope within 20 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal, cytoplasmic domain of mouse NR2B.

Application

Anti-NR2B, Cat. No. 06-600, is a rabbit polyclonal antibody that detects NR2B and is tested for use in Western Blotting.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neurodegenerative Diseases

Quality

Evaluated by Western Blotting in rat brain microsomal preparation. Western Blotting Analysis: A 1:1,000 dilution of this antibody detected NR2B in rat brain microsomal preparation.

Target description

Target molecular weight ~165 kDa observed; 165.96 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Store at -10°C to -25°C. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance

Analysis Note

Control
Mouse brain tissue

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perturbing PSD-95 interactions with NR2B-subtype receptors attenuates spinal nociceptive plasticity and neuropathic pain.
D'Mello, R; Marchand, F; Pezet, S; McMahon, SB; Dickenson, AH
Molecular Therapy null
Duncan Sinclair et al.
Journal of neurodevelopmental disorders, 8, 14-14 (2016-05-03)
Neurodevelopmental disorders such as autism spectrum disorders and schizophrenia differentially impact males and females and are highly heritable. The ways in which sex and genetic vulnerability influence the pathogenesis of these disorders are not clearly understood. The n-methyl-d-aspartate (NMDA) receptor
Peripheral N-methyl-d-aspartate receptors contribute to mechanical hypersensitivity in a rat model of inflammatory temporomandibular joint pain.
Ivanusic, JJ; Beaini, D; Hatch, RJ; Staikopoulos, V; Sessle, BJ; Jennings, EA
European Journal of Pain null
Amino terminal domain regulation of NMDA receptor function.
Herin, Greta Ann and Aizenman, Elias
European Journal of Pharmacology, 500, 101-111 (2004)
N-methyl-D-aspartate receptor subunit NR3a expression and function in principal cells of the collecting duct.
Sproul, A; Steele, SL; Thai, TL; Yu, S; Klein, JD; Sands, JM; Bell, PD
American Journal of Physiology: Renal Physiology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service