Skip to Content
Merck
All Photos(1)

Key Documents

05-652

Sigma-Aldrich

Anti-MUC1/episialin Antibody, clone 214D4

clone 214D4, Upstate®, from mouse

Synonym(s):

Breast carcinoma-associated antigen DF3, Carcinoma-associated mucin, Episialin, H23AG, PEMT, Peanut-reactive urinary mucin, PUM, Polymorphic epithelial mucin, PEM, Tumor-associated epithelial membrane antigen, EMA, Tumor-associated mucin, CD227

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

clone

214D4, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

MUC1/episialin

Immunogen

Fusion protein, human b-gal-episialin/MUC1 repeat domain

Application

Anti-MUC1/episialin Antibody, clone 214D4 detects level of MUC1/episialin & has been published & validated for use in FC, IH.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

routinely evaluated by immunohistochemistry

Target description

none given

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilene K Gipson et al.
PloS one, 9(6), e100393-e100393 (2014-06-27)
Membrane-anchored mucins are present in the apical surface glycocalyx of mucosal epithelial cells, each mucosal epithelium having at least two of the mucins. The mucins have been ascribed barrier functions, but direct comparisons of their functions within the same epithelium
A mechanism for inhibition of E-cadherin-mediated cell-cell adhesion by the membrane-associated mucin episialin/MUC1.
Wesseling, J, et al.
Molecular Biology of the Cell, 7, 565-577 (1996)
Pablo Argüeso et al.
The Journal of biological chemistry, 284(34), 23037-23045 (2009-06-27)
Maintenance of an intact mucosal barrier is critical to preventing damage to and infection of wet-surfaced epithelia. The mechanism of defense has been the subject of much investigation, and there is evidence now implicating O-glycosylated mucins on the epithelial cell
Monoclonal antibodies against the nonmucin domain of MUC1/episialin.
Hilkens, J and Boer, M
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 19 Suppl 1, 67-70 (1998)
Y Tatematsu et al.
Bone marrow transplantation, 47(3), 416-425 (2011-05-17)
The ocular surface is a frequent target tissue of mucosal chronic GVHD (cGVHD). We investigated the histopathological features of the conjunctival microvilli in patients with cGVHD. Conjunctival tissue specimens from patients with cGVHD or Sjögren's syndrome (SS) or from healthy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service