Skip to Content
Merck
All Photos(2)

Key Documents

05-532

Sigma-Aldrich

Anti-CaM Kinase II Antibody, α subunit, clone 6G9

clone 6G9, Upstate®, from mouse

Synonym(s):

Anti-Anti-CAMKA, Anti-Anti-CaMKIINalpha, Anti-Anti-CaMKIIalpha, Anti-Anti-MRD53, Anti-Anti-MRT63

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G9, monoclonal

species reactivity

rat, bovine

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... CAMK2A(815)

Specificity

α subunit of CaM Kinase II

Immunogen

Partially purified rat CaM Kinase II, α-subunit

Application

Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling
This Anti-CaM Kinase II Antibody, α subunit, clone 6G9 is validated for use in IH, WB for the detection of CaM Kinase II.

Quality

routinely evaluated by immunoblot on bovine brain cytosol or rat brain microsome preparations

Target description

50kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% azide before the addition of glycerol to 30%
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raquel V Fornari et al.
Frontiers in behavioral neuroscience, 6, 10-10 (2012-03-22)
Glucocorticoids are known to enhance the consolidation of memory of emotionally arousing experiences by acting upon a network of interconnected brain regions. Although animal studies typically do not consider the insular cortex (IC) to be part of this network, the
C C Huang et al.
The Journal of biological chemistry, 276(51), 48108-48117 (2001-10-27)
Reversal of long term potentiation (LTP) may function to increase the flexibility and storage capacity of neuronal circuits; however, the underlying mechanisms remain incompletely understood. We show that depotentiation induced by low frequency stimulation (LFS) (2 Hz, 10 min, 1200
E M Stanley et al.
Neuroscience, 195, 70-79 (2011-09-03)
GABAergic neurons of the medial septum of the basal forebrain make up a substantial portion of the septo-hippocampal pathway fibers, and are known to modulate hippocampal amino acid neurotransmission and support cognitive function. Importantly, these neurons are also implicated in
Ca(2+)/calmodulin-dependent protein kinase II inhibition by heparin in mesangial cells.
Xiao, W; Liu, Y; Templeton, DM
American Journal of Physiology: Renal Physiology null
Synergistic activation of dopamine D1 and TrkB receptors mediate gain control of synaptic plasticity in the basolateral amygdala.
Li, C; Dabrowska, J; Hazra, R; Rainnie, DG
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service