Skip to Content
Merck
All Photos(2)

Key Documents

05-480

Sigma-Aldrich

Anti-Eck/EphA2 Antibody, clone D7

clone D7, Upstate®, from mouse

Synonym(s):

EPH receptor A2, Epithelial cell kinase, Tyrosine-protein kinase receptor ECK, ephrin receptor EphA2, epithelial cell receptor protein tyrosine kinase, protein tyrosine kinase, receptor protein tyrosine kinase regulated by p53 and E2F-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

D7, monoclonal

species reactivity

mouse, canine, human, rat, bovine

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

bovine ... Epha2(512798)
human ... EPHA2(1969)

General description

EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands.
Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation.

Specificity

This antibody recognizes Epithelial Cell Kinase (Eck/EphA2), Mr 140 kDa

Immunogen

Native protein isolated by purification of phosphotyrosine-containing proteins. Clone D7.

Application

Protein Kinase Assay:
A previous lot of this antibody has been reported to have been used in an immunoprecipitation autophosphorylation assay, using a Mn-PIPES reaction buffer (Romer, L., 1994).

Immunoprecipitation:
A previous lot of this antibody has been reported to immunoprecipitate Eck from 500 µg of a human breast epithelial cell line which had been lysed in TBS containing 1% Triton X-100. Use 1-4 µg per reaction.

Immunocytochemistry:
A previous lot of this antibody has been reported to immunostain Eck in human, mouse and rat epithelial cells fixed with 3.7% formaldehyde solution and permeabilized with 0.5% Triton X-100 in TBS.
Research Category
Neuroscience
Research Sub Category
Growth Cones & Axon Guidance
This Anti-Eck/EphA2 Antibody, clone D7 is validated for use in IC, IP, EA, WB for the detection of Eck/EphA2.

Quality

Routinely evaluated by immunoblot on RIPA lysates from human A431, foreskin fibroblasts, murine 3T3/A31 or rat L6 cells.

Western Blot Analysis:
0.5-2 µg/mL of this lot detected Eck in RIPA lysates from human A431 and previously from foreskin fibroblasts, murine 3T3/A31 and rat L6 cells.

Target description

140 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1 in buffer containing 0.2 M Tris-glycine, pH 7.4, 0.15 M NaCl, with 0.05% sodium azide and 30% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations:
Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. NOTE: Variability in freezer temperatures below -20°C may cause glycerol-containing solutions to become frozen during storage.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Magal et al.
Journal of neuroscience research, 43(6), 735-744 (1996-03-15)
Although the Eph subfamily represents the largest group of receptor protein-tyrosine kinases, the biological roles of the Eph-related receptors and their ligands are not well understood. B61 has been identified recently by receptor affinity chromatography as a ligand for the
Christine Gundry et al.
Nature communications, 8, 14646-14646 (2017-03-16)
The Rab GTPase effector, Rab-coupling protein (RCP) is known to promote invasive behaviour in vitro by controlling integrin and receptor tyrosine kinase (RTK) trafficking, but how RCP influences metastasis in vivo is unclear. Here we identify an RTK of the
Predictive value of the EphA2 receptor tyrosine kinase in lung cancer recurrence and survival.
Michael S Kinch, Mary-Beth Moore, David H Harpole
Clinical cancer research : an official journal of the American Association for Cancer Research null
A R Hess et al.
Cancer research, 61(8), 3250-3255 (2001-04-20)
During embryogenesis, blood vessels are formed initially by the process of vasculogenesis, the in situ differentiation of mesenchymal cells into endothelial cells, which form a primitive, patterned vasculogenic network. This is followed by angiogenesis, the sprouting of new vessels from
Identification of tyrosine phosphorylated adhesion proteins in human cancer cells.
Kinch, M S, et al.
Hybridoma, 17, 227-235 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service