Skip to Content
Merck
All Photos(5)

Documents

03-102

Sigma-Aldrich

RIPAb+ HuR - RIP Validated Antibody and Primer Set

from rabbit

Synonym(s):

ELAV-like protein 1, Hu-antigen R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

biological source

rabbit

Quality Level

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

RIPAb+
Upstate®

technique(s)

RIP: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... ELAVL1(1994)

General description

RIPAb+ antibodies are evaluated using the RNA Binding Protein Immunoprecipitation (RIP) assay. Each RIPAb+ antibody set includes quantitative RT-PCR control primers (RIP Primers) to biologically validate your IP results by successfully coprecipitating a specific RNA target (where such a specific target is known). The qPCR protocol and primer sequences are provided, allowing researchers to validate RIP protocols when using the antibody in their experimental context. Each set also includes a negative control antibody to ensure specificity of the RIP reaction.
The RIPAb+ HuR set includes the HuR antibody, a negative control antibody (purified Rabbit IgG), and positive qPCR primers which amplify a 100 bp fragment of the human beta actin (ACTB) cDNA. The HuR and negative control antibodies are supplied in a scalable "per RIP" reaction size and can be used to functionally validate the precipitation HuR-associated RNAs.

Immunogen

Epitope: a.a. 1-13 of HuR

Application

Immunoprecipitation from RIP lysate:
RIP lysate from MCF7 cells (2 X 106 cell equivalents per IP) was subjected to immunoprecipitation using 0.5 μg of either a normal rabbit IgG or Anti-HuR antibody. Precipitated proteins were resolved by electrophoresis, transferred to nitrocellulose and probed with anti-HuR (1.0 μg/mL).
Proteins were visualized using One-Step IP-Western kit (GenScript Cat. # L00231) (Please see figures).

Western Blot Analysis:
3T3/A31 lysate was resolved by electrophoresis, transferred to PVDF membranes and probed with HuR, hu (1:500 dilution). Proteins were visualized using a Donkey anti-Rabbit conjugated to HRP and visualized using a chemiluminescence detection system. (Please see figures).

Immunoprecipitation Analysis:
10 ug of this antibody immunoprecipitated HuR from 500 ug of 3T3/A31 RIPA lysate (Please see figures).

mmunocytochemistry Analysis:
Confocal IF analysis of HeLa, NIH/3T3 using anti-HuR (Red). Actin filaments have been labeled with AlexaFluor 488 -Phalloidin (Green). Nucleus is stained with DAPI (Blue) (Please see figures).
This RIPAb+ HuR -RIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Quality

RNA binding protein Immunoprecipitation:
RIP Lysate prepared from HeLa cells (2 x 107 cell equivalents per IP) were subjected to immunoprecipitation using 5 µg of either a normal Rabbit IgG or Anti-HuR antibody and the Magna RIP Kit (Cat. # 17-700).
Successful immunoprecipitation of HuR-associated RNA was verified by qPCR using RIP Primers, ACTB (Please see figures).
Please refer to the Magna RIP® (Cat. # 17-700) or EZ-Magna RIP (Cat. # 17-701) protocol for experimental details.

Target description

35 kDa

Physical form

Anti-HuR (rabbit polyclonal). One vial containing 50 μg of purified rabbit polyclonal in 100 μL of buffer containing PBS in 0.05% sodium azide with 50% Glycerol. Liquid at -20°C.

Normal Rabbit IgG. One vial containing 125 μg purified rabbit IgG in 125 μL storage buffer containing 0.1% sodium azide. Store at -20°C.

RIP Primers, ACTB. One vial containing 75 μL of 5 μM of each primer specific for human beta actin (ACTB). Store at -20°C.
FOR: TTG TTA CAG GAA GTC CCT TGC C
REV: ATG CTA TCA CCT CCC CTG TGT G
Format: Purified

Analysis Note

Control
Included negative control rabbit IgG antibody and control primers specific for human beta actin (ATCB).

Legal Information

MAGNA RIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Pu et al.
Frontiers in oncology, 11, 754835-754835 (2021-11-05)
Hepatocellular carcinoma (HCC) is one of the most aggressive malignancies. Increasing evidence revealed that long noncoding RNAs (lncRNAs) were frequently involved in various malignancies. Here, we explored the clinical significances, roles, and mechanisms of lncRNA ADORA2A antisense RNA 1 (ADORA2A-AS1)
Luana Carvalho et al.
Molecular psychiatry (2023-08-04)
We previously discovered using transcriptomics that rats undergoing withdrawal after chronic ethanol exposure had increased expression of several genes encoding RNA splicing factors in the hippocampus. Here, we examined RNA splicing in the rat hippocampus during withdrawal from chronic ethanol
Heat resistance of Bacillus spores when adhered to stainless steel and its relationship to spore hydrophobicity.
P Simmonds,B L Mossel,T Intaraphan,H C Deeth
Journal of Food Protection null
Xiangbo Ruan et al.
Nature communications, 11(1), 45-45 (2020-01-04)
Unlike protein-coding genes, the majority of human long non-coding RNAs (lncRNAs) are considered non-conserved. Although lncRNAs have been shown to function in diverse pathophysiological processes in mice, it remains largely unknown whether human lncRNAs have such in vivo functions. Here
Jesús Morillo-Bernal et al.
Cancers, 16(2) (2024-01-23)
RNA-binding proteins play diverse roles in cancer, influencing various facets of the disease, including proliferation, apoptosis, angiogenesis, senescence, invasion, epithelial-mesenchymal transition (EMT), and metastasis. HuR, a known RBP, is recognized for stabilizing mRNAs containing AU-rich elements (AREs), although its complete

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service