Skip to Content
Merck
All Photos(1)

Documents

Z-001

Supelco

Zolpidem-D6 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15D6N3O
CAS Number:
Molecular Weight:
313.43
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule B (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

suitability

suitable for (Not suitable for use with GC/MS)

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CC1=CC=C(C=C1)C2=C(CC(N(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)N3C=C(C)C=CC3=N2

InChI

1S/C19H21N3O/c1-13-5-8-15(9-6-13)19-16(11-18(23)21(3)4)22-12-14(2)7-10-17(22)20-19/h5-10,12H,11H2,1-4H3/i3D3,4D3

InChI key

ZAFYATHCZYHLPB-LIJFRPJRSA-N

General description

Zolpidem is a non-benzodiazepine hypnotic sold under the trade names Ambien®, Stilnox® and Edluar® for the treatment of insomnia. This stable-labeled internal standard is suitable for quantitation of zolpidem levels in urine, serum, or plasma by LC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods. Zolpidem has a wide history of abuse, including recreational use and as a date-rape drug.

Legal Information

Ambien is a registered trademark of Sanofi S.A.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Edluar is a registered trademark of Meda Pharmaceuticals Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Stilnox is a registered trademark of Sanofi S.A.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service