Skip to Content
Merck
All Photos(1)

Key Documents

N-077

Supelco

(S)-N-Nitrosoanabasine (NAB) solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

NAB, NAB solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N3O
CAS Number:
Molecular Weight:
191.23
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=NN1CCCC[C@H]1c2cccnc2

InChI

1S/C10H13N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h3-4,6,8,10H,1-2,5,7H2/t10-/m0/s1

InChI key

BXYPVKMROLGXJI-JTQLQIEISA-N

General description

N-Nitrosoanabasine or NAB is a potent carcinogen and biomarker of exposure to cigarette smoke. It′s found in a variety of tobacco products including chewing tobacco and snuff. Certified Spiking Solutions® are suitable for use in LC-MS/MS or GC/MS methods for nicotine testing, environmental monitoring, or clinical and diagnostic testing of nicotine biomarkers.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTIONS is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irina Stepanov et al.
Journal of agricultural and food chemistry, 50(10), 2793-2797 (2002-05-02)
Tobacco-specific nitrosamines (TSNA) are among the most important and abundant strongly carcinogenic agents in unburned tobacco. It has been established that 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) induces lung tumors in rodents independent of the route of administration. N'-Nitrosonornicotine (NNN) causes tumors of the
A M Idris et al.
Carcinogenesis, 12(6), 1115-1118 (1991-06-01)
The aim of these studies was to determine the levels of carcinogenic tobacco specific nitrosamines (TSNA) in Sudanese oral snuff (toombak) as recent retrospective epidemiological studies suggested an association between the use of toombak and subsequent development of oral cancer.
B Hasséus et al.
European journal of oral sciences, 105(1), 45-51 (1997-02-01)
Smokeless tobacco (ST) is known to adversely effect the oral mucosa, but knowledge about the influence on immune defence is limited. Few studies have investigated the effect of ST on the local immune response. In the present study, we have
B G Osterdahl
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 28(9), 619-622 (1990-09-01)
In many countries nicotine-containing chewing gum (Nicorette) is used to help to break the habit of smoking. Saliva was collected every 5 min from chewers of nicotine chewing gum and analysed for tobacco-specific nitrosamines. Detectable levels of tobacco-specific nitrosamines were
A M Idris et al.
Carcinogenesis, 13(6), 1001-1005 (1992-06-01)
Exposure to tobacco-specific nitrosamines (TSNA) has been measured in the saliva of 12 users of Sudanese oral snuff (toombak). Using GC coupled to thermal energy analysis, levels of N'-nitrosonornicotine (NNN), N'-nitrosoanatabine (NAT), N'-nitrosoanabasine (NAB) and 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone (NNK) were measured before

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service