Skip to Content
Merck
All Photos(1)

Key Documents

N-017

Supelco

Normeperidine solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO2
CAS Number:
Molecular Weight:
233.31
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(OCC)C1(C2=CC=CC=C2)CCNCC1

InChI

1S/C14H19NO2/c1-2-17-13(16)14(8-10-15-11-9-14)12-6-4-3-5-7-12/h3-7,15H,2,8-11H2,1H3

InChI key

QKHMFBKXTNQCTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Normeperidine is a toxic primary metabolite of the opioid analgesic meperidine. This Certified Snap-N-Spike® Solution is suitable for use in pain prescription monitoring, clinical toxicology, urine drug testing, or forensic analysis by LC-MS/MS, LC/MS or GC/MS. Meperidine, a synthetic opioid with effects similar to morphine, is used as a painkiller.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Plummer et al.
Toxicology, 95(1-3), 37-44 (1995-01-06)
This study investigated behavioural effects of the toxic pethidine metabolite, norpethidine, in rats and its interactions with reserpine, apomorphine and physostigmine. Following intraperitoneal administration, brain concentrations of norpethidine reached a plateau after 20-40 min and remained elevated for 2 h.
Thomas T Simopoulos et al.
Archives of surgery (Chicago, Ill. : 1960), 137(1), 84-88 (2002-01-31)
Intravenous patient-controlled analgesia (IV PCA) meperidine hydrochloride can be used with a reasonable margin of safety. A retrospective review was performed of 355 medical records of patients receiving IV PCA meperidine treatment. Four groups of patients were defined, based on
Meperidine in patient-controlled analgesia: a near-fatal mishap.
R J Geller
Anesthesia and analgesia, 76(3), 655-657 (1993-03-01)
J H Vranken et al.
Acta anaesthesiologica Scandinavica, 49(5), 665-670 (2005-04-20)
Intrathecal administration of meperidine, an opioid with local anesthetic activity, can induce analgesia in patients with intractable cancer pain. However, continuous intrathecal administration may result in the accumulation of normeperidine, responsible for central nervous system toxicity. Ten patients with neuropathic
E Nissen et al.
Acta paediatrica (Oslo, Norway : 1992), 86(2), 201-208 (1997-02-01)
A standard dose of 100 mg of pethidine was given im to 13 healthy primiparae during labour. The aim of the study was to investigate whether developing breastfeeding behaviour in the newborn infant was associated with the dose-delivery time interval

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service