Skip to Content
Merck
All Photos(1)

Documents

M-144

Supelco

MDAI hydrochloride solution

100 μg/mL in acetonitrile: water (9:1) with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2 · HCl
CAS Number:
Molecular Weight:
213.66
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in acetonitrile: water (9:1) with 5% 1 M HCl (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−70°C

SMILES string

Cl.NC1Cc2cc3OCOc3cc2C1

InChI

1S/C10H11NO2.ClH/c11-8-1-6-3-9-10(13-5-12-9)4-7(6)2-8;/h3-4,8H,1-2,5,11H2;1H

InChI key

DEZYWEZDXRXACY-UHFFFAOYSA-N

General description

MDAI, or 5,6-Methylenedioxy-2-aminoindane, is a new stimulant and club/designer drug with significant cases of abuse reported in Asia, Europe and the US. Sold over the Internet as a “bath salt,” MDAI is a potential successor to mephedrone as the next legal high drug.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M P Johnson et al.
Pharmacology, biochemistry, and behavior, 40(4), 915-922 (1991-12-01)
There is increasing evidence linking dopamine (DA) to the long-term serotonergic (5-HT) neurotoxic effects of certain substituted amphetamines such as 3,4-methylenedioxymethamphetamine (MDMA). The present study was undertaken to examine the importance of DA metabolism, uptake inhibition and release in the
R Oberlender et al.
The Journal of pharmacology and experimental therapeutics, 255(3), 1098-1106 (1990-12-01)
The stimulus properties of 3,4-methylenedioxymethamphetamine (MDMA)-like compounds were studied in rats trained to discriminate saline from (+)-N-methyl-1-(1,3-benzodioxol-5-yl)-2-butanamine [(+)-MBDB] hydrochloride (7.18 mumol/kg; 1.75 mg/kg), the alpha-ethyl homolog of MDMA. In previous experiments with (+)-MBDB as a test drug, complete substitution was
Cathal T Gallagher et al.
Human psychopharmacology, 27(2), 106-112 (2012-03-06)
The fully synthetic 'legal high' 5,6-methylenedioxy-2-aminoindane (MDAI) is an analogue of 3,4-methylenedioxymethamphetamine. Although developed in the 1990s, it was not widely abused until 2010. However, mephedrone was banned in the UK in April 2010, and almost immediately, MDAI was widely
Laurent Karila et al.
Drug testing and analysis, 3(9), 552-559 (2011-10-01)
Designer drugs belong to a group of legally or illegally produced substances that are structurally and pharmacologically very similar to illicit drugs. In the past, designer drugs were often used during all-night dance parties, but they are now consumed in
Roland P Archer et al.
Drug testing and analysis, 3(7-8), 505-514 (2011-07-12)
Historically, the appearance of new psychoactive materials (and hence the requirement for new reference standards) has been relatively slow. This position has now changed, with 101 new psychoactive substances reported to EMCDDA-Europol since 2006. The newly reported materials, and associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service