I-030
Interference Mix Kit (For Qualitative Use Only)
analytical standard, Cerilliant®
Synonym(s):
Qualitative Interference Mixture
About This Item
Recommended Products
grade
analytical standard
Quality Level
form
liquid
manufacturer/tradename
Cerilliant®
drug control
estupefaciente/psicótropo (Spain); Decreto Lei 15/93: Tabela IA/IIB/IV (Portugal)
technique(s)
gas chromatography (GC): suitable
liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
multi-component solution
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- Screening for cellular aging mechanisms: An Interference Mix Kit was employed to explore the senescence of hematopoietic stem cells in vitro, assisting in the understanding of cellular memory and aging processes, crucial for advancements in regenerative medicine (Dong et al., 2021).
- Enhanced diagnostic assays for SARS-CoV-2: The kit was utilized in the evaluation of RT-qPCR mastermixes for detecting SARS-CoV-2 RNA, demonstrating its application in enhancing the reliability of diagnostic tests during the COVID-19 pandemic (Haddar et al., 2020).
- Contact sensitivity in chronic wounds: This kit facilitated a multicentric study to evaluate contact sensitivity in patients with venous leg ulcers, proving essential for identifying allergenic compounds and improving treatment strategies (Rai et al., 2018).
- Forensic species identification: Utilized in a collaborative inter-laboratory exercise, the Interference Mix Kit aided in validating the SPInDel approach for species identification, crucial for forensic science applications (Alves et al., 2017).
- Detection of Brucella spp. in dairy: The kit supported the development of real-time quantitative PCR for screening Brucella spp. in bovine raw milk, highlighting its role in ensuring food safety and public health (Kaynak-Onurdag et al., 2016).
Packaging
Legal Information
Kit Components Also Available Separately
- I-023Interference Mix 1 (For Qualitative Use Only), 100-1,000 μg/mL each component in methanol, analytical standard, Cerilliant® 1 mLSDS
- I-024Interference Mix 2 (For Qualitative Use Only), 100 μg/μL each analyte in methanol, analytical standard, Cerilliant® 1 mLSDS
- I-025Interference Mix 3 (For Qualitative Use Only), 50-1000 μg/mL each component in acetonitrile, analytical standard, Cerilliant® 1 mLSDS
- I-026Interference Mix 4 (For Qualitative Use Only), 5-100 μg/mL each component in methanol, Cerilliant® 1 mLSDS
- I-027Interference Mix 5, 100 μg/mL in methanol (each analyte), Cerilliant® 1 mLSDS
- I-028Interference Mix 6, 5-100 μg/mL each component in methanol, Cerilliant® 1 mLSDS
- I-029Interference Mix 7 (For Qualitative Use Only), 50-100 μg/mL in methanol (each analyte), Cerilliant® 1 mLSDS
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1A - STOT RE 2 - STOT SE 1
Target Organs
Blood, Eyes,Central nervous system
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service