Skip to Content
Merck
All Photos(1)

Key Documents

H-027

Supelco

(±)-11-Hydroxy-Δ9-THC solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O3
CAS Number:
Molecular Weight:
330.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

psicótropo (Spain); Decreto Lei 15/93: Tabela IIB (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

cannabis testing
forensics and toxicology

format

single component solution

storage temp.

2-8°C

SMILES string

OC1=CC(CCCCC)=CC2=C1C3C=C(CO)CCC3C(O2)(C)C

InChI

1S/C21H30O3/c1-4-5-6-7-14-11-18(23)20-16-10-15(13-22)8-9-17(16)21(2,3)24-19(20)12-14/h10-12,16-17,22-23H,4-9,13H2,1-3H3

InChI key

YCBKSSAWEUDACY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® applicable in LC/MS or GC/MS applications for cannabinoid testing, urine drug testing, clinical toxicology, or forensic analysis. 11-Hydroxy-Δ9-THC is the primary oxidation metabolite of the main psychoactive component of cannabis, Δ9-Tetrahydrocannabinol (Δ9-THC).

Application

The Certified Spiking Solution® is suitable for (±)-11-Hydroxy-Δ9-THC testing by HPLC, GC/MS, or LC-MS/MS methods in forensic analysis, clinical toxicology, urine drug testing, and Cannabis potency or impurity profiling testing applications. The Certified Reference Material (CRM) in solution can also be used as follows:
  • Quantification of cannabidiol and Δ9-tetrahydrocannabinol in addition to the identification of minor cannabinoids from human plasma samples using ultra-performance liquid chromatography (UHPLC) combined with triple quadrupole mass spectrometry following a liquid-liquid extraction
  • Liquid chromatography-tandem mass spectrometry (LC-MS/MS) method-based determination of 10 cannabinoids in oral fluid samples following their solid phase extraction (SPE)
  • Analysis of Δ9-Tetrahydrocannabinol (Δ9-THC) and its three metabolites in plasma and urine samples by gas chromatography (GC) coupled to triple quadrupole mass spectrometer
  • Development of an automated sample preparation method based on solid phase extraction (SPE) to determine ∆9-tetrahydrocannabinol (THC) and its three primary metabolites in a variety of matrices, such as serum, saliva, and urine using liquid chromatography-mass spectrometry (LC-MS)
  • Simultaneous determination of cannabinoids and their metabolites in human serum samples by an atmospheric-pressure chemical ionization gas chromatography-tandem mass spectrometry (APGC-MS/MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cynthia Coulter et al.
Journal of analytical toxicology, 32(8), 653-658 (2008-11-15)
An analytical procedure for the determination of Delta9-tetrahydrocannabinol (THC), 11-nor-9-carboxy-Delta9-tetrahydrocannabinol (THCA), and 11-hydroxy-Delta9-tetrahydrocannabinol (11-OH-THC) in whole blood has been developed and validated using liquid chromatography with tandem mass spectral detection (MS). Cannabinoids present in the blood samples were quantified using
Erin L Karschner et al.
Clinical chemistry, 57(1), 66-75 (2010-11-17)
Sativex(®), a cannabis extract oromucosal spray containing Δ(9)-tetrahydrocannabinol (THC) and cannabidiol (CBD), is currently in phase III trials as an adjunct to opioids for cancer pain treatment, and recently received United Kingdom approval for treatment of spasticity. There are indications
Erin L Karschner et al.
Analytical and bioanalytical chemistry, 397(2), 603-611 (2010-03-23)
A sensitive analytical method for simultaneous quantification of sub-nanogram concentrations of cannabidiol (CBD), Delta(9)-tetrahydrocannabinol (THC), 11-hydroxy-THC (11-OH-THC), and 11-nor-9-carboxy-THC (THCCOOH) in plasma is presented for monitoring cannabinoid pharmacotherapy and illicit cannabis use. Analytes were extracted from 1 mL plasma by
J Röhrich et al.
Journal of analytical toxicology, 34(4), 196-203 (2010-05-15)
Cannabinoid concentrations in blood and urine after passive exposure to cannabis smoke under real-life conditions were investigated in this study. Eight healthy volunteers were exposed to cannabis smoke for 3 h in a well-attended coffee shop in Maastricht, Netherlands. An
Stefan W Toennes et al.
Archiv fur Kriminologie, 225(3-4), 90-98 (2010-05-29)
When defendants are confronted with evidence of cannabinoids in their blood suggesting consumption of cannabis they sometimes argue that this could only be due to a passive exposure. The small number of controlled studies available showed that tetrahydrocannabinol (THC), the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service