Skip to Content
Merck
All Photos(1)

Key Documents

F-031

Supelco

Fluconazole solution

2.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12F2N6O
CAS Number:
Molecular Weight:
306.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

2.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Fc1c(ccc(c1)F)C(O)(C[n]3ncnc3)C[n]2ncnc2

InChI

1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22H,4-5H2

InChI key

RFHAOTPXVQNOHP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fluconazole, sold under trade names such as Trican, Monicure, Diflucan®, and Afungil, is an antifungal drug used in the treatment and prevention of superficial and systemic fungal infections. This Certified Spiking Solution® is suitable as starting material for calibrators, controls, or linearity standards for therapeutic drug monitoring or clinical and diagnostic testing of fluconazole in patient blood, serum, or plasma samples by HPLC or LC-MS/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Diflucan is a registered trademark of Pfizer, Inc.
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Pfaller et al.
Drug resistance updates : reviews and commentaries in antimicrobial and anticancer chemotherapy, 13(6), 180-195 (2010-11-06)
Both the Clinical and Laboratory Standards Institute (CLSI) and the European Committee on Antimicrobial Susceptibility Testing (EUCAST) have MIC clinical breakpoints (CBPs) for fluconazole (FLU) and Candida. EUCAST CBPs are species-specific, and apply only to C. albicans, C. tropicalis and
Mahipal Sinnollareddy et al.
Expert opinion on drug metabolism & toxicology, 7(11), 1431-1440 (2011-09-03)
Invasive candidiasis has emerged over the last few decades as an increasingly important nosocomial problem for the critically ill, affecting around 2% of intensive care unit patients. Although poor outcomes associated with invasive candidiasis among critically ill patients may relate
Rajendra J Kothavade et al.
Journal of medical microbiology, 59(Pt 8), 873-880 (2010-04-24)
Candida tropicalis has been identified as the most prevalent pathogenic yeast species of the Candida-non-albicans group. Historically, Candida albicans has been the major species responsible for causing candidiasis in immunocompromised and immunocompetent patients. However, infections (candidiasis) due to C. tropicalis
Jayantha Thiyanaratnam et al.
Journal of drugs in dermatology : JDD, 9(10), 1272-1275 (2010-10-15)
Fluconazole uncommonly causes Stevens-Johnson syndrome. A young Indian man who developed this adverse effect after his second dose of fluconazole is described. The characteristics of previously reported individuals with fluconazole-associated Stevens-Johnson syndrome and toxic epidermal necrolysis are also summarized: three
Raymond A Smego et al.
Medicine, 90(4), 237-249 (2011-06-23)
The treatment of Candida infective endocarditis generally involves infected valve removal accompanied by antifungal therapy with amphotericin B or a lipid-based derivative, with or without flucytosine. While often used as chronic suppressive therapy in these patients, the precise role for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service