Skip to Content
Merck
All Photos(1)

Documents

F-001

Supelco

Fentanyl-D5 solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22D5H23N2O
CAS Number:
Molecular Weight:
341.50
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

-10 to -25°C

SMILES string

[2H]c1c([2H])c([2H])c(c([2H])c1[2H])N(C2CCN(CC2)CCc3ccccc3)C(=O)CC

InChI

1S/C22H28N2O/c1-2-22(25)24(20-11-7-4-8-12-20)21-14-17-23(18-15-21)16-13-19-9-5-3-6-10-19/h3-12,21H,2,13-18H2,1H3/i4D,7D,8D,11D,12D

InChI key

PJMPHNIQZUBGLI-SCUSKTIWSA-N

General description

An internal standard suitable for fentanyl testing or isotope dilution methods by GC/MS or LC/MS for clinical toxicology, urine drug testing, pain prescription monitoring, or forensic analysis. Fentanyl is a potent synthetic narcotic analgesic and local anesthetic sold under various trade names including Sublimaze, Duregesic, Actiq®, and Fentora®.

Application


  • Fentanyl-D₅ analytical standard: Stability and compatibility of an intramuscular fetal anesthetic cocktail, which includes Fentanyl-D₅, were studied for fetal intervention. This highlights Fentanyl-D₅′s role as a reliable analytical standard in the preparation of precise medication dosages essential for sensitive applications (Ross et al., 2024).

  • Opioid research chemical: Fentanyl-D₅ was used in developing a method for quantifying nitazene analogs in Dried Blood Spots using liquid chromatography-tandem mass spectrometry, underlining its importance as a research chemical in the study of synthetic opioids (Ververi et al., 2024).

  • Mass spectrometry Fentanyl: The determination of nine fentanyl drugs in hair samples by GC-MS/MS and LC-MS/MS employed Fentanyl-D₅, showcasing its critical application in forensic toxicology and pharmacokinetic studies to track opioid exposure (Wei and Su, 2022).

  • Synthetic opioid calibration solution: A novel bioanalytical method determined opioids in blood and pericardial fluid, utilizing Fentanyl-D₅ as part of the calibration solution. This study highlights its pivotal role in enhancing the reliability of quantitative toxicological analysis (Ferreira et al., 2020).

Legal Information

Actiq is a registered trademark of Anesta Corp.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Fentora is a registered trademark of Cima Labs, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Shen et al.
Biochemical and biophysical research communications, 464(3), 794-799 (2015-07-19)
Paraquat (PQ) was demonstrated to induce dopaminergic neuron death and is used as a Parkinson's disease (PD) mimetic. Amounting evidences demonstrated that Rho/ROCK may a novel target for the therapy of PD. Previously we synthesized L-F001 and proved it is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service