Skip to Content
Merck
All Photos(1)

Key Documents

E-116

Supelco

Ethyl sulfate sodium salt

10 mg/mL in methanol: water (1:1) ((as ethyl sulfate)), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5O4SNa
CAS Number:
Molecular Weight:
148.11
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

10 mg/mL in methanol: water (1:1) ((as ethyl sulfate))

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

[Na+].CCOS([O-])(=O)=O

InChI

1S/C2H6O4S.Na/c1-2-6-7(3,4)5;/h2H2,1H3,(H,3,4,5);/q;+1/p-1

InChI key

RRLOOYQHUHGIRJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, or urine alcohol testing. Ethyl sulfate is a primary urinary metabolite of consumed alcohol and is indicative of recent alcohol consumption.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This study is aimed at developing and validating an LC-ESI-ion trap-MS/MS method for identification and quantification of ethyl glucuronide (EtG) and ethyl sulfate (EtS) as ethanol biomarkers for urine analysis.

This study is aimed at developing and validating an LC-ESI-ion trap-MS/MS method for identification and quantification of ethyl glucuronide (EtG) and ethyl sulfate (EtS) as ethanol biomarkers for urine analysis.

This study is aimed at developing and validating an LC-ESI-ion trap-MS/MS method for identification and quantification of ethyl glucuronide (EtG) and ethyl sulfate (EtS) as ethanol biomarkers for urine analysis.

This study is aimed at developing and validating an LC-ESI-ion trap-MS/MS method for identification and quantification of ethyl glucuronide (EtG) and ethyl sulfate (EtS) as ethanol biomarkers for urine analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service