Skip to Content
Merck
All Photos(1)

Documents

C-028

Supelco

R(+)-Cathinone hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12ClNO
CAS Number:
Molecular Weight:
185.65
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

Cl.C[C@@H](N)C(=O)c1ccccc1

InChI

1S/C9H11NO.ClH/c1-7(10)9(11)8-5-3-2-4-6-8;/h2-7H,10H2,1H3;1H/t7-;/m1./s1

InChI key

HPZCAKYHISJOIK-OGFXRTJISA-N

General description

Cathinone, or benzoylethanamine, is a major active constituent of the recreational drug “Khat”. This stimulant has also been found in illicit bath salts. This Certified Snap-N-Spike® Solution is suitable for use in LC-MS/MS or GC/MS testing applications such as urine drug testing, clinical toxicology, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S E Shortall et al.
British journal of pharmacology, 168(4), 966-977 (2012-10-10)
Recreational users report that mephedrone has similar psychoactive effects to 3,4-methylenedioxymethamphetamine (MDMA). MDMA induces well-characterized changes in body temperature due to complex monoaminergic effects on central thermoregulation, peripheral blood flow and thermogenesis, but there are little preclinical data on the
Przemysław Biliński et al.
Annals of agricultural and environmental medicine : AAEM, 19(4), 857-870 (2013-01-15)
Faced with the rapidly growing increase of designer drug abuse, particularly amongst the younger generation, various legislative strategies are currently employed world-wide for tackling this problem - however with mixed results. The key issue is that the producers of DDs
John D Power et al.
Rapid communications in mass spectrometry : RCM, 26(22), 2601-2611 (2012-10-13)
Amphetamine-like cathinone derivatives have become popular as recreational drugs over the past several years but their identification for forensic purposes is made difficult as they undergo extensive fragmentation under commonly used electron ionization (EI) conditions to afford ambiguous mass spectra.
Psychoactive 'bath salts': compounds, mechanisms, and toxicities.
Kurt R Lehner et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 38(1), 243-244 (2012-11-14)
Jassim Al Suwaidi et al.
Clinica chimica acta; international journal of clinical chemistry, 419, 11-14 (2013-02-02)
This article reviews current knowledge in khat (cathinone) research and its health impacts and toxicity in the cardiac system of khat chewers based on current evidence. The authors describe the process by which khat is postulated to induce multiple cardiac

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service