Skip to Content
Merck
All Photos(1)

Key Documents

B-044

Supelco

Buprenorphine solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H41NO4
CAS Number:
Molecular Weight:
467.64
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIC (Portugal)

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

CO[C@]12CC[C@@]3(C[C@@H]1[C@](C)(O)C(C)(C)C)[C@H]4Cc5ccc(O)c6O[C@@H]2[C@]3(CCN4CC7CC7)c56

InChI

1S/C29H41NO4/c1-25(2,3)26(4,32)20-15-27-10-11-29(20,33-5)24-28(27)12-13-30(16-17-6-7-17)21(27)14-18-8-9-19(31)23(34-24)22(18)28/h8-9,17,20-21,24,31-32H,6-7,10-16H2,1-5H3/t20-,21-,24-,26+,27-,28+,29-/m1/s1

InChI key

RMRJXGBAOAMLHD-IHFGGWKQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

A Certified Spiking Solution® suitable for GC/MS or LC/MS applications in pain prescription monitoring, forensic testing, or clinical toxicology.

Buprenorphine is a semi-synthetic opioid that shows partial agonistic activity towards the μ-opioid receptor and antagonistic activity towards the kappa opioid receptor. And even though it is similar to morphine, it is more potent than morphine. The drug is sold under numerous trade names such as Suboxone®, Subutex®, and Temgesic.

Application

The certified reference solution (CRM) in solution can also be used as follows:
  • Multi-residue analysis of buprenorphine, norbuprenorphine, and naloxone in the fingernails and urine samples of previous heroin users under an opioid substitution therapy by solid-liquid or solid-phase extraction combined with liquid chromatography-mass spectrometry (LC-MS) for analysis
  • Liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS) based separation and determination of naloxone, buprenorphine, and norbuprenorphine in human plasma samples following their single-step liquid-liquid extraction (LLE)
  • Development of nanocomposite rhodium nanoparticles-mesoporous carbon (RhNPs-MC) for their deposition on glassy carbon electrode (GCE) to compose an electrochemical sensor for the measurement of morphine and buprenorphine in pharmaceutical formulations and human serum samples using differential pulse voltammetry (DPV)
  • Study of human blood plasma samples for the quantitative analysis of buprenorphine and naloxone by liquid chromatography (LC) coupled to high-resolution time-of-flight mass spectrometry (HR-Q-TOF-MS)
  • Simultaneous determination of buprenorphine, norbuprenorphine, and their glucuronides from human urine samples by ultra-high-performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Suboxone is a registered trademark of RB Pharmaceuticals Limited
Subutex is a registered trademark of RB Pharmaceuticals Limited

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johan Menten et al.
Cancer investigation, 31(6), 412-420 (2013-06-14)
Many head and neck cancer (HNC) patients experience painful therapy-related mucositis and dermatitis. This prospective observational study evaluated transdermal buprenorphine use in HNC patients to relieve treatment-related pain. During treatment with paracetamol or tramadol, visual analogue scale (VAS)-pain scores >30/100
Karen L Cropsey et al.
Journal of addiction medicine, 7(3), 210-215 (2013-04-24)
This project sought to demonstrate the feasibility and acceptability of providing on-site buprenorphine treatment to individuals under community corrections supervision. Seventeen women and 13 men were enrolled on-site over a 2-week period at a community corrections location. Study participants received
The clinical conundrum of perioperative pain management in patients with opioid dependence: lessons from two cases.
Jacqueline S Israel et al.
Plastic and reconstructive surgery, 131(4), 657e-658e (2013-04-02)
K Wahle et al.
MMW Fortschritte der Medizin, 155 Suppl 1, 25-31 (2013-05-18)
Chronic pain in the elderly is common. Especially in the elderly inadequate treatment of pain can cause significant functional impairmentand deterioration of qualityof life. Theaim of this post-marketing surveillance study was to collect data from clinical practice on the analgesic
Jatinder Mohan Chawla et al.
Journal of opioid management, 9(1), 35-41 (2013-05-28)
Tramadol is a synthetic opiate and a centrally acting weak m-opioid receptor agonist. The potential advantages of tramadol include ease of administration, low abuse potential, and being nonscheduled. This study compared tramadol and buprenorphine for controlling withdrawal symptoms in patients

Articles

Although both biphenyl and phenyl-hexyl phases can resolve these compounds, the former exhibits excellent peak shape and substantially less silanol-derived ion exchange activity.

Protocols

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

Optimize β-glucuronidase hydrolysis for glucuronide metabolite analysis considering factors like time, temperature, pH, and enzyme concentration.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service