Skip to Content
Merck
All Photos(1)

Documents

B-022

Supelco

Bufotenine solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16N2O
CAS Number:
Molecular Weight:
204.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule D (Switzerland); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

OC1=CC=C(NC=C2CCN(C)C)C2=C1

InChI

1S/C12H16N2O/c1-14(2)6-5-9-8-13-12-4-3-10(15)7-11(9)12/h3-4,7-8,13,15H,5-6H2,1-2H3

InChI key

VTTONGPRPXSUTJ-UHFFFAOYSA-N

General description

Bufotenine is a hallucinogenic alkaloid found in mushrooms, plants, mammals and in toads belonging to the Bufo genus. This drug is structurally similar to psilocin, a psychedelic compound found in mushrooms. This Snap-N-Spike® Reference Solution is suitable for use in urine drug testing, clinical toxicology, or forensic analysis by LC-MS/MS or GC/MS.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T L Barry et al.
Journal of forensic sciences, 41(6), 1068-1073 (1996-11-01)
The death of a 23-year-old man resulting from digoxin-like toxicity and heart failure was attributed to ingestion of a West Indian aphrodisiac known as "Love Stone." GC/MS analyses identified bufotenine, a controlled substance under both US and New York State
Barbara J Ebersole et al.
Molecular pharmacology, 63(1), 36-43 (2002-12-19)
Based on experiment and computational simulation, we present a structural explanation for the differing efficacies of indole agonists at the human serotonin 5-HT2A receptor (5HT2AR). We find that serotonin [5-hydroxytryptamine (5-HT)] forms hydrogen-bonds with Ser3.36 in helix 3 and Ser5.46
T O G Costa et al.
Toxicon : official journal of the International Society on Toxinology, 46(4), 371-375 (2005-08-02)
Bufotenin (5-hydroxy-N,N-dimetyltryptamine) is a tryptamine alkaloid widely spread among anuran families as a component of their chemical defense system, acting as a potent hallucinogenic factor, showing similar activity to LSD upon interaction with the 5HT2 human receptor. This work demonstrates
B L Roth et al.
Molecular pharmacology, 52(2), 259-266 (1997-08-01)
Several models of agonist binding to G protein-coupled 5-hydroxytryptamine [5-HT] (serotonin) receptors have highlighted the potential importance of highly conserved aromatic residues for ligand binding and agonist efficacy. In this study, we tested these models by constructing and characterizing a
K A Berg et al.
Molecular pharmacology, 54(1), 94-104 (1998-07-11)
There are many examples of a single receptor coupling directly to more than one cellular signal transduction pathway. Although traditional receptor theory allows for activation of multiple cellular effectors by agonists, it predicts that the relative degree of activation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service