Skip to Content
Merck
All Photos(1)

Documents

A-915

Supelco

7-Aminoclonazepam solution

100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12ClN3O
CAS Number:
Molecular Weight:
285.73
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

NC1=CC=C2C(C(C3=C(Cl)C=CC=C3)=NCC(N2)=O)=C1

InChI

1S/C15H12ClN3O/c16-12-4-2-1-3-10(12)15-11-7-9(17)5-6-13(11)19-14(20)8-18-15/h1-7H,8,17H2,(H,19,20)

InChI key

HEFRPWRJTGLSSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified Snap-N-Spike® solution is suitable for use as a starting material in calibrators and controls for numerous GC/MS or LC/MS applications in forensic analysis, clinical toxicology and urine drug testing. As a major metabolite, 7-aminoclonazepam may be used to monitor use of the parent drug, clonazepam. Clonazepam, marketed as Klonopin® and Rivotril, is a long-acting benzodiazepine with anxiolytic, anticonvulsant, muscle relaxant, and hypnotic properties.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Klonopin is a registered trademark of Hoffmann-La Roche Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marjorie Chèze et al.
Forensic science international, 145(2-3), 123-130 (2004-09-29)
The number of reports on drug facilitated crimes is increasing these last years. Apart from ethanol and cannabis, benzodiazepines (BZD) and analogs are the most common drugs reported to be used probably due to their amnesic and sedative properties. We
D R Peng et al.
Pediatric pharmacology (New York, N.Y.), 4(3), 155-159 (1984-01-01)
The N-acetylation of the reduced metabolite of clonazepam 7-amino-clonazepam was studied in cytosolic preparation from human fetal and adult livers. The metabolite formed 7-acetamido-clonazepam was measured with high performance liquid chromatography. A bimodal distribution of the N-acetyltransferase activities was observed
Wei Chen et al.
Biosensors & bioelectronics, 24(7), 2051-2056 (2008-12-09)
The present paper described a rapid and ultrasensitive detection method using a microfluidic chip for analyzing 7-aminoclonazepam (7-ACZP) residues in human urine. A microfluidic chip-based immunoassay with laser-induced fluorescence (LIF) detection based on the water-soluble denatured bovine serum albumin (dBSA)-coated
A Negrusz et al.
Journal of analytical toxicology, 24(7), 614-620 (2000-10-24)
Clonazepam (CLO) is an anticonvulsant benzodiazepine approved by the Food and Drug Administration for use in the treatment of seizures. It produces pharmacological effects (depression, amnesia) similar to other compounds from the same therapeutic class, and in combination with alcohol
Simple high-performance liquid chromatographic assay for the routine monitoring of clonazepam in plasma.
R L Heazlewood et al.
Journal of chromatography, 336(1), 229-233 (1984-12-07)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service