Skip to Content
Merck
All Photos(1)

Key Documents

A-034

Supelco

Anhydroecgonine methyl ester solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO2
CAS Number:
Molecular Weight:
181.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®. SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule A (Switzerland)

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

COC(=O)C1=CCC2CCC1N2C

InChI

1S/C10H15NO2/c1-11-7-3-5-8(10(12)13-2)9(11)6-4-7/h5,7,9H,3-4,6H2,1-2H3/t7-,9-/m1/s1

InChI key

MPSNEAHFGOEKBI-VXNVDRBHSA-N

General description

Anhydroecgonine methyl ester, or methylecgonidine, is a metabolite of smoked crack cocaine. This cocaine analog is also used as an ingredient in the production of phenyltropanes, a class of drugs used to treat cocaine dependency. This certified solution standard is suitable for use in LC/MS or GC/MS applications for urine drug testing, clinical toxicology, or forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hilary James Liberty et al.
Addiction biology, 8(2), 191-200 (2003-07-10)
A continuing social problem is presented by the large number of individuals who use crack cocaine. Recent research has identified unique pyrolysis products of crack or burned cocaine as anhydroecgonine methylester (AEME) and ecgonidine (ECD) through gas chromatography/mass spectrometry (GC/MS)
Anabel S Fandiño et al.
Chemical research in toxicology, 15(12), 1543-1548 (2002-12-17)
During smoking of cocaine-base (crack), anhydroecgonine methyl ester (AEME, methylecgonidine) is formed in large amounts as a pyrolysis product of cocaine and is absorbed in the lungs. The metabolism of AEME was studied in the present investigation using microsome preparations
Detection of smoked cocaine marker (anhydroecgonine methylester) in nails.
C Ragoucy-Sengler et al.
Journal of analytical toxicology, 29(7), 765-768 (2006-01-20)
Karl B Scheidweiler et al.
The Journal of pharmacology and experimental therapeutics, 307(3), 1179-1187 (2003-10-17)
Methylecgonidine is formed from cocaine base when smoked and has been identified in biological fluids of crack smokers. Ecgonidine, a metabolite of methylecgonidine formed via esterase activity, also has been identified in similar samples collected from crack smokers. Methylecgonidine and
Anabel S Fandiño et al.
Journal of analytical toxicology, 26(8), 567-570 (2002-12-28)
The presence of the cocaine pyrolysis product anhydroecgonine methyl ester (AEME, methylecgonidine) in plasma indicates the smoking of cocaine. The stability of this analyte in human plasma has not been studied. In the present investigation AEME and its hydrolysis product

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service