Skip to Content
Merck
All Photos(1)

Key Documents

860585P

Avanti

17:0 SM (d18:1/17:0)

Avanti Research - A Croda Brand 860585P, powder

Synonym(s):

N-heptadecanoyl-D-erythro-sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H81N2O6P
CAS Number:
Molecular Weight:
717.05
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860585P-10mg)
pkg of 1 × 5 mg (860585P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860585P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C40H81N2O6P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-40(44)41-38(37-48-49(45,46)47-36-35-42(3,4)5)39(43)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h31,33,38-39,43H,6-30,32,34-37H2,1-5H3,(H-,41,44,45,46)/b33-31+/t38-,39+/m0/s1

InChI key

YMQZQHIESOAPQH-JXGHDCMNSA-N

Application

N-heptadecanoyl-D-erythro-sphingosylphosphorylcholine (17:0 SM (d18:1/17:0)) has been used as an internal standard for the quantification of C17-sphingomyelin (SM) using mass spectrometry (MS). It may be used as an internal standard for the quantification of lipids in serum and skeletal muscles using liquid chromatography-mass spectrometry (LC-MS) analysis. It may also be used as an internal standard for retention time calibration and for the extraction of lipids from blood samples using methanol and methyl tert-butyl ether (MTBE).

Packaging

5 mL Amber Glass Screw Cap Vial (860585P-10mg)
5 mL Amber Glass Screw Cap Vial (860585P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianing Mi et al.
Journal of agricultural and food chemistry, 66(34), 8991-8998 (2018-07-31)
Deep profiling of glycosphingolipids and sphingomyelins in wild Cordyceps was carried out by using offline chromatographic enrichment followed by ultrahigh performance liquid chromatography-ultrahigh definition-quadrupole time-of-flight mass spectrometry (UHPLC-UHD-Q-TOF-MS). A total of 119 glycosphingolipids (72 new ones) and 87 sphingomyelins (43
Huili Zhang et al.
Translational psychiatry, 10(1), 138-138 (2020-05-14)
Fluoxetine is a commonly prescribed antidepressant, and the mechanisms of increased bone fragility with its long-term use remain largely unknown. Here, we show that long-term administration of fluoxetine induces the disruption of sphingolipids metabolism in bone marrow adipose tissue (BMAT)through
Peng Wu et al.
Chinese medicine, 15, 9-9 (2020-01-31)
Our previous clinical evidence suggested that the direct application of "Sanse powder" the main ingredient of "Yiceng" might represent an alternative treatment for knee osteoarthritis. However, the mechanism underlying its effect is poorly understood. In this study, we investigated the
Dinesh Kumar Barupal et al.
Scientific data, 5, 180263-180263 (2018-11-21)
Alzheimer's disease (AD) is a major public health priority with a large socioeconomic burden and complex etiology. The Alzheimer Disease Metabolomics Consortium (ADMC) and the Alzheimer Disease Neuroimaging Initiative (ADNI) aim to gain new biological insights in the disease etiology.
Ryutaro Adachi et al.
European journal of medicinal chemistry, 136, 283-293 (2017-05-16)
Sphingomyelin synthase (SMS) is a membrane enzyme that catalyzes the synthesis of sphingomyelin, is required for the maintenance of plasma membrane microdomain fluidity, and has two isoforms: SMS1 and SMS2. Although these isoforms exhibit the same SMS activity, they are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service