Skip to Content
Merck
All Photos(1)

Key Documents

Z743745

ART® Low Retention SoftFit L Reload System

HLT 1200L, size 1200 μL, reload insert, non-sterile

Synonym(s):

aerosol barrier tips, barrier filter pipette tip, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121603
NACRES:
NB.02
manufacturer/tradename:
Molecular BioProducts 3742-05-RI
size:
1200 μL
sterility:
non-sterile
feature:
HLT 1200L
reload insert
packaging:
pkg of 4X768 ea

sterility

non-sterile

feature

HLT 1200L
reload insert

packaging

pkg of 4X768 ea

manufacturer/tradename

Molecular BioProducts 3742-05-RI

technique(s)

PCR: suitable

size

1200 μL

Looking for similar products? Visit Product Comparison Guide

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service