Skip to Content
Merck
All Photos(1)

Key Documents

Z743727

ART® Low Retention Barrier Tip

ART 10 REACH, size 10 μL, racked, sterile

Synonym(s):

aerosol barrier tips, barrier filter tips, barrier tip, filter pipette tip, filter tip, pipet tip, sterile barrier pipette tip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121603
NACRES:
NB.02
volume:
10 μL
sterility:
sterile
feature:
ART 10 REACH
racked
packaging:
pkg of 5X960 ea
manufacturer/tradename:
Molecular BioProducts 2140-05

sterility

sterile

feature

ART 10 REACH
racked

packaging

pkg of 5X960 ea

manufacturer/tradename

Molecular BioProducts 2140-05

technique(s)

PCR: suitable

size

10 μL

volume

10 μL

Looking for similar products? Visit Product Comparison Guide

General description

  • ART® low retention barrier tips are designed to maximize sample recovery.
  • Tip′s hydrophobic inner surface prevents sample loss, facilitates sample accuracy and reproducibility.
  • Clear tips provide sample clarity.
  • Self-sealing technology prevents cross-contamination.
  • Certified RNase, DNase, ATP, endotoxin, and pyrogen free.
ART™ Barrier Reload System with low retention tips helps to perform applications precisely and consistently. This system ensures both accuracy and protection while maintaining sterility. With the ability to stack tips due to offset wells, it minimizes waste and optimizes benchtop and storage space.

Features and Benefits

  • ART low retention barrier tips ensure complete protection against aerosols and liquid contamination.
  • The tips are free from DNase, RNase, DNA, ATP, PCR inhibitors, and endotoxins.
  • Stackable barrier tip packaging enables tips to pass through offset holes in the lower insert.
  • Blister sleeve guide rails facilitate smooth transfer of tip inserts into the rack.
  • An outer sleeve shields tips from external contamination during the refilling of inserts.
  • The tip packaging option helps reduce plastic waste.
  • The rigid insert design ensures reliable tip loading.
  • Lot number and description are printed on every insert for easy tracking and identification.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service