Z263206
Wheaton wide-mouth bottle
capacity 16 oz (500 mL), white polypropylene cap, clear, vinyl flat liner
Synonym(s):
Wheaton bottle, Wheaton bottles, wide mouth glass
About This Item
Recommended Products
material
round bottle
soda-lime glass bottle
vinyl flat liner
white polypropylene cap
cap size
63-400
Joint
Joints threaded neck (63-400)
sterility
non-sterile
feature
graduations: no
with cap
with cap
packaging
pkg of 24 ea
manufacturer/tradename
WHEATON® W216926
capacity
16 oz (500 mL)
diam. × H
79 mm × 133 mm
color
clear
Looking for similar products? Visit Product Comparison Guide
General description
- Wheaton wide-mouth bottle is manufactured from type III soda-lime glass.
- Clear bottles for optimal sample clarity.
- Ideal for storing and transporting small to large volume samples or dry powders.
- A wide-mouth design facilitates easy access to samples.
- Includes a tamper-evident seal for sample protection.
Features and Benefits
- Available in larger bottle sizes
- Wide mouth / neck
- Clear, Type III soda-lime glass
- Available with caps attached or without caps
- HAZCOM labels, provided in each case of bottles with the exception of bulk packs, allow for ID of content and safety notification
- Tamper Evident Seal keeps bottle sealed, protecting contents from accidental misuse (not included in bulk packs)
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service