Skip to Content
Merck
All Photos(4)

Key Documents

W6934

L-Lactic acid-1-13C

greener alternative

API for Clinical Studies, ≥99 atom % 13C, ≥95% (CP)

Synonym(s):

(S)-2-Hydroxypropionic acid-1-13C, Sarcolactic acid-1-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)13CO2H
CAS Number:
Molecular Weight:
91.07
MDL number:
UNSPSC Code:
51113400

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

grade

cGMP

isotopic purity

≥99 atom % 13C

Assay

≥95% (CP)

greener alternative product score

old score: 45
new score: 14
Find out more about DOZN™ Scoring

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Design for Energy Efficiency
Inherently Safer Chemistry for Accident Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

mass shift

M+1

storage temp.

-10 to -25°C

SMILES string

C[C@H](O)[13C](O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1/i3+1

InChI key

JVTAAEKCZFNVCJ-NSQKCYGPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses”, “Design for Energy Efficiency” and “Inherently Safer Chemistry for Accident Prevention”. Click here to view its DOZN scorecard.

Packaging

Typically, this product is available to ship immediately from stock. Contact Stable Isotopes Customer Service to discuss quantity and packaging needs.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service