Skip to Content
Merck
All Photos(1)

Key Documents

W518905

Sigma-Aldrich

Sodium propionate

FCC

Synonym(s):

Propionic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2COONa
CAS Number:
Molecular Weight:
96.06
Beilstein:
3566934
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.21
grade:
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Halal
Kosher

reg. compliance

FCC
FDA 21 CFR 150.141
FDA 21 CFR 150.161
FDA 21 CFR 179.45
FDA 21 CFR 180.23
FDA 21 CFR 184.1784

Assay

99%

mp

285-286 °C (lit.)

solubility

H2O: 10%, clear, colorless

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

faint

SMILES string

[Na+].CCC([O-])=O

InChI

1S/C3H6O2.Na/c1-2-3(4)5;/h2H2,1H3,(H,4,5);/q;+1/p-1

InChI key

JXKPEJDQGNYQSM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Dipentene is one of the flavor constituents of spearmint oil. It used in toothpaste and electronic cigarettes.[1][2]
Sodium propionate is generally used as a preservative in bread due to its anti-microbial and anti-mycotic properties.[3][4]

Application


  • The Effects of Short-Chain Fatty Acids in Gut Immune and Oxidative Responses of European Sea Bass (Dicentrarchus labrax): An Ex Vivo Approach.: This study examines the beneficial effects of sodium propionate on gut health and immune responses in European sea bass, highlighting its potential as a dietary supplement to enhance fish health and disease resistance (Fontinha F et al., 2024).

  • Melatonin protects zebrafish pancreatic development and physiological rhythms from sodium propionate-induced disturbances via the hypothalamic-pituitary-thyroid axis.: Research investigates how sodium propionate impacts endocrine functions in zebrafish, suggesting its broader implications in endocrinology and toxicology studies (Xu Y et al., 2024).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effect of Flavoring Chemicals on Free Radical Formation in Electronic Cigarette Aerosols.
Bitzer ZT, et al.
Free Radical Biology & Medicine, 112, 200-200 (2017)
Evaluation of genotoxic effects of sodium propionate, calcium propionate and potassium propionate on the root meristem cells of Allium cepa.
Turko?lu S.
Food And Chemical Toxicology, 46(6), 2035-2041 (2008)
Contact allergy to toothpaste flavors.
Andersen KE.
Contact Dermatitis, 4(4), 195-198 (1978)
Comparative analysis of the antimicrobial activity of cinnamon oil and cinnamon extract on some food-borne microbes.
Gupta C, et al.
African Journal of Microbiology Research, 2(9), 247-251 (2008)
Bistra B Nankova et al.
PloS one, 9(8), e103740-e103740 (2014-08-30)
Alterations in gut microbiome composition have an emerging role in health and disease including brain function and behavior. Short chain fatty acids (SCFA) like propionic (PPA), and butyric acid (BA), which are present in diet and are fermentation products of

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service