Skip to Content
Merck
All Photos(1)

Key Documents

W512303

Sigma-Aldrich

Glutaric dialdehyde solution

50 wt. % in H2O, FCC

Synonym(s):

Glutaraldehyde solution, Glutaric dialdehyde solution, Pentane-1,5-dial

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OHC(CH2)3CHO
CAS Number:
Molecular Weight:
100.12
Beilstein:
605390
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
Flavis number:
5.149
NACRES:
NA.21
grade:
Kosher
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Kosher

reg. compliance

FCC

form

liquid

concentration

50 wt. % in H2O
50.0-52.5% (FCC monograph)

impurities

may contain ≤0.5% Methanol (residual solvent)

refractive index

n20/D 1.42

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

medicinal; pungent

SMILES string

[H]C(CCCC([H])=O)=O

InChI

1S/C5H8O2/c6-4-2-1-3-5-7/h4-5H,1-3H2

InChI key

SXRSQZLOMIGNAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Synthesis and Characterization of N-Isopropylacrylamide Microspheres as pH Sensors.: This article details the development of microspheres from glutaric dialdehyde solution for use as pH sensors, showcasing their potential in biochemistry applications for sensitive and accurate pH monitoring (Lavine et al., 2021).

  • Characterization and application of chondroitin sulfate/polyvinyl alcohol nanofibres prepared by electrospinning.: This study investigates the properties of chondroitin sulfate/polyvinyl alcohol nanofibers crosslinked with glutaric dialdehyde. The fibers exhibit enhanced biocompatibility and structural integrity, suggesting their utility in advanced drug delivery systems and tissue engineering (Guo et al., 2016).

  • Thermosensitive Behavior and Antibacterial Activity of Cotton Fabric Modified with a Chitosan-poly(N-isopropylacrylamide) Interpenetrating Polymer Network Hydrogel.: This publication presents the modification of cotton fabric with a hydrogel formed using glutaric dialdehyde. The fabric demonstrates significant thermosensitive and antibacterial properties, suitable for developing smart textiles and wound dressings (Wang et al., 2016).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphane Jauréguiberry et al.
Blood, 124(2), 167-175 (2014-05-27)
Patients with severe malaria treated with artesunate sometimes experience a delayed hemolytic episode. Artesunate (AS) induces pitting, a splenic process whereby dead parasites are expelled from their host erythrocytes. These once-infected erythrocytes then return to the circulation. We analyzed hematologic
Shoichi Kageyama et al.
Liver transplantation : official publication of the American Association for the Study of Liver Diseases and the International Liver Transplantation Society, 21(1), 112-122 (2014-09-23)
Preconditioning by brief ischemia protects not only the concerned organ but also other distant organs against subsequent lethal damage; this is called remote ischemic preconditioning (RIPC). This study was designed to investigate the impact of intestinal RIPC on hepatic ischemia/reperfusion
Stephanie J Bissel et al.
Journal of neuropathology and experimental neurology, 74(8), 767-777 (2015-06-27)
Human parechovirus 3 (HPeV3) is a picornavirus associated with neurologic disease in neonates. Human parechovirus 3 infection of preterm and term infants is associated with seizures and destructive periventricular white matter lesions. Despite unremarkable cerebrospinal fluid (CSF), HPeV3 RNA can
Irene Costantini et al.
Scientific reports, 5, 9808-9808 (2015-05-08)
Extensive mapping of neuronal connections in the central nervous system requires high-throughput µm-scale imaging of large volumes. In recent years, different approaches have been developed to overcome the limitations due to tissue light scattering. These methods are generally developed to
Andri K Riau et al.
Investigative ophthalmology & visual science, 55(5), 3186-3194 (2014-04-26)
To compare flap adhesion strength, stromal bed quality, and tissue responses after flap preparation using nJ- and μJ-energy level femtosecond lasers. All corneal flaps were created by either VisuMax laser (μJ-energy level) or femto-LDV (nJ-energy level). Flap adhesion strength in

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service