Skip to Content
Merck
All Photos(1)

Key Documents

W447704

Sigma-Aldrich

Ethyl 2-methyl-1,3-dioxolane-2-acetate

≥99%

Synonym(s):

Fructone, apple Ketal, 2-Methyl-1,3-dioxolane-2-acetic acid ethyl ester, Applinal, Ethyl 2-methyldioxolan-2-yl acetate, Jasmaprunat, NSC 6537

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
FEMA Number:
4477
Beilstein:
130164
MDL number:
UNSPSC Code:
12164502
NACRES:
NA.21
Organoleptic:
apple; fruity; sweet
grade:
Fragrance grade
Halal
Kosher
biological source:
synthetic
Agency:
follows IFRA guidelines
meets purity specifications of JECFA
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

grade

Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009

Assay

≥99%

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

apple; fruity; sweet

SMILES string

O=C(OCC)CC1(C)OCCO1

InChI

1S/C8H14O4/c1-3-10-7(9)6-8(2)11-4-5-12-8/h3-6H2,1-2H3

InChI key

XWEOGMYZFCHQNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

197.6 °F - Pensky-Martens closed cup

Flash Point(C)

92 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service