Skip to Content
Merck
All Photos(1)

Key Documents

W382108

Sigma-Aldrich

3-Carene

≥90%

Synonym(s):

δ3-Carene, 3,7,7-Trimethylbicyclo[4.1.0]hept-3-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
FEMA Number:
3821
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
1.029
NACRES:
NA.21
biological source:
synthetic
food allergen:
no known allergens
Pricing and availability is not currently available.

biological source

synthetic

Assay

≥90%

refractive index

n20/D 1.474 (lit.)

bp

168-169 °C/705 mmHg (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

citrus; sweet

SMILES string

CC1=CCC2C(C1)C2(C)C

InChI

1S/C10H16/c1-7-4-5-8-9(6-7)10(8,2)3/h4,8-9H,5-6H2,1-3H3

InChI key

BQOFWKZOCNGFEC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Carene is a volatile monoterpene compound typically found in essential oils such as turpentine oil[1], juniper oil[2] and pine needle oil.[3]

Application

  • Identification and quality evaluation of Lushan Yunwu tea from different geographical origins based on metabolomics.: The presence of 3-carene in Lushan Yunwu tea is studied for its impact on tea quality and flavor profile, providing insights into the metabolomic distinctions influenced by geographical factors (Sun et al., 2024).

  • Influence of Maqian essential oil on gut microbiota and immunoresponses in type 1 diabetes: In silico study.: This in silico study investigates how 3-carene, as a component of Maqian essential oil, influences the gut microbiota and immune responses in type 1 diabetes, suggesting potential therapeutic uses of 3-carene in managing autoimmune diseases (Dahab et al., 2024).

Disclaimer

For R&D or non-EU Food use. Not for retail sale.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F - closed cup

Flash Point(C)

47 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemical composition of the essential oils of Juniperus from ripe and unripe berries and leaves and their antimicrobial activity.
Angioni A, et al.
Journal of Agricultural and Food Chemistry, 51(10), 3073-3078 (2003)
Studies on chemical constituents of the volatile oil from the needle of Pinus elliottii Engelm.
Chen H, et al.
Natural Product Research and Development, 13(3), 36-38 (2001)
Selected oxidized fragrance terpenes are common contact allergens.
Matura M, et al.
Contact Dermatitis, 52(6), 320-328 (2005)
Yan Ma et al.
Physical chemistry chemical physics : PCCP, 11(21), 4184-4197 (2009-05-22)
This paper describes experimental studies aimed at elucidating mechanisms for the formation of low-volatility organic acids in the gas-phase ozonolysis of 3-carene. Experiments were carried out in a static chamber under 'OH-free' conditions. A range of multifunctional acids-which are analogous
Ta-Jung Lu et al.
The Journal of organic chemistry, 76(6), 1621-1633 (2011-02-11)
A novel carene-based alanine-equivalent tricyclic iminolactone 16 has been synthesized via stereoselective dihydroxylation of the double bond, IBX oxidation of the secondary alcohol, esterification of the tertiary alcohol, deprotection of the resulting ester, and subsequent cyclization from commercially available (1S)-(+)-3-carene

Protocols

Fast GC analysis of sweet orange essential oil in hexane. Key components identified includes: β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service