Skip to Content
Merck
All Photos(1)

Documents

W379905

Sigma-Aldrich

1,2-Dimethoxybenzene

≥99%, FG

Synonym(s):

Pyrocatechol dimethyl ether, Veratrole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OCH3)2
CAS Number:
Molecular Weight:
138.16
FEMA Number:
3799
Beilstein:
1364621
EC Number:
Council of Europe no.:
10320
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
4.062
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002

Assay

≥99%

refractive index

n20/D 1.533 (lit.)

bp

206-207 °C (lit.)

mp

15 °C (lit.)

density

1.084 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

anise; clove; spicy; vanilla

SMILES string

COc1ccccc1OC

InChI

1S/C8H10O2/c1-9-7-5-3-4-6-8(7)10-2/h3-6H,1-2H3

InChI key

ABDKAPXRBAPSQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Vincent J Chebny et al.
Organic letters, 11(11), 2253-2256 (2009-05-29)
Definitive X-ray crystallographic evidence is obtained for a single hole (or a polaron) to be uniformly distributed on the three equivalent 1,2-dimethoxybenzenoid (or veratrole) rings in the hexamethoxytriptycene cation radical. This conclusion is further supported by electrochemical analysis and by
Umma Salma Jhumur et al.
Journal of medical entomology, 43(6), 1164-1170 (2006-12-14)
Flower odors are important signals for chemical communication between plants and flower visitors. Here, we studied the naive responses of Culex pipiens pipiens biotype molestus Forskal 1775 (Diptera: Culicidae) to typical flower odors and assessed the learning capacity of mosquitoes
D Cai et al.
The Journal of biological chemistry, 267(16), 11149-11155 (1992-06-05)
The present study characterizes the serial reactions of H2O2 with compounds I and II of lignin peroxidase isozyme H1. These two reactions constitute part of the pathway leading to formation of the oxy complex (compound III) from the ferric enzyme.
Yoshiya Inokuchi et al.
Physical chemistry chemical physics : PCCP, 14(13), 4457-4462 (2012-02-23)
We report UV photodissociation (UVPD) and IR-UV double-resonance spectra of 1,2-dimethoxybenzene (DMB) complexes with alkali metal ions, M(+)·DMB (M = Li, Na, K, Rb, and Cs), in a cold, 22-pole ion trap. The UVPD spectrum of the Li(+) complex shows
Sandra Kreher et al.
Molecular microbiology, 78(1), 230-237 (2010-10-07)
The anaerobic cleavage of ether bonds of methoxylated substrates such as vanillate or veratrol in acetogenic bacteria is mediated by multi-component enzyme systems, the O-demethylases. Acetobacterium dehalogenans harbours different inducible O-demethylases with various substrate spectra. Two of these enzyme systems

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service