Skip to Content
Merck
All Photos(1)

Key Documents

W329703

Sigma-Aldrich

L-Leucine

99%, FG

Synonym(s):

(S)-2-Amino-4-methylpentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
FEMA Number:
3297
Beilstein:
1721722
EC Number:
Council of Europe no.:
10482
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
17.012
NACRES:
NA.21

biological source

plant

Quality Level

grade

FG
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.320
FDA 21 CFR 172.804

Assay

99%

optical activity

[α]20/D +15.5°, c = 2 in 6 M HCl

mp

>300 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

odorless

SMILES string

CC(C)C[C@H](N)C(O)=O

InChI

1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1

InChI key

ROHFNLRQFUQHCH-YFKPBYRVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Leucine is a branched-chain amino acid mainly found in whey proteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Whey proteins in the regulation of food intake and satiety.
Luhovyy BL, et al.
Journal of the American College of Nutrition, 26(6), 704S-712S (2007)
Irena Roci et al.
Cell reports, 26(7), 1691-1700 (2019-02-14)
Alterations in cell-cycle regulation and cellular metabolism are associated with cancer transformation, and enzymes active in the committed cell-cycle phase may represent vulnerabilities of cancer cells. Here, we map metabolic events in the G1 and SG2M phases by combining cell
T Yoshimasa et al.
The Journal of clinical investigation, 69(3), 643-650 (1982-03-01)
To elucidate the physiological and pathophysiological significance of methionine- and leucine-enkephalin (Met-and Leu-enkephalin, respectively) in human sympathoadrenal system, the contents of these peptides in normal human sympathetic nervous system, adrenal medulla, and pheochromocytomas were determined by specific radioimmunoassays combined with
Gaetano Santulli et al.
The Journal of clinical investigation, 125(5), 1968-1978 (2015-04-07)
The type 2 ryanodine receptor (RyR2) is a Ca2+ release channel on the endoplasmic reticulum (ER) of several types of cells, including cardiomyocytes and pancreatic β cells. In cardiomyocytes, RyR2-dependent Ca2+ release is critical for excitation-contraction coupling; however, a functional
Marika Leenders et al.
Nutrition reviews, 69(11), 675-689 (2011-10-28)
Amino acids function as precursors for de novo protein synthesis. In addition, however, they play a key role as nutritional signals that regulate multiple cellular processes. There is ample in vitro and in vivo evidence showing that muscle tissue responds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service