Skip to Content
Merck
All Photos(1)

Key Documents

W313505

Sigma-Aldrich

trans,trans-2,4-Decadienal

≥90%, FG

Synonym(s):

(E,E)-2,4-Decadienal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH=CHCHO
CAS Number:
Molecular Weight:
152.23
FEMA Number:
3135
Beilstein:
1704897
EC Number:
Council of Europe no.:
2120
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
5.140
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

Agency

meets purity specifications of JECFA

reg. compliance

EU Regulation 1334/2008 & 178/2002

vapor density

>1 (vs air)

Assay

≥90%

refractive index

n20/D 1.515 (lit.)

bp

114-116 °C/10 mmHg (lit.)

density

0.872 g/mL at 20 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; meaty; citrus

SMILES string

[H]C(=O)\C=C\C=C\CCCCC

InChI

1S/C10H16O/c1-2-3-4-5-6-7-8-9-10-11/h6-10H,2-5H2,1H3/b7-6+,9-8+

InChI key

JZQKTMZYLHNFPL-BLHCBFLLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans,trans-2,4-Decadienal is one of the main volatile flavor constituents of rice, tomato, cold-pressed orange and tangerine oils. It has also been identified in frying oils and fumes since it is formed as a by-product when lipids in edible oils undergo peroxidation on heating.

Application


  • Multispectral and Molecular Dynamics Study on the Impact of trans,trans-2,4-Decadienal and 4-hydroxy-2-nonenal on Myoglobin Redox Stability.: The study examines the effects of lipid oxidation products on protein stability, with significant implications for understanding oxidative stress and its impact on muscle foods (Qi et al., 2024).

Packaging

Packaged in glass bottles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F - closed cup

Flash Point(C)

101 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Content of trans, trans?2, 4?decadienal in deep?fried and pan?fried potatoes.
Boskou G, et al.
European Journal of Lipid Science and Technology, 108(2), 109-115 (2006)
Flavour compounds and a quantitative descriptive analysis of tomatoes (Lycopersicon esculentum Mill.) of different cultivars in short-term storage.
Krumbein A, et al.
Postharvest biology and technology, 32(1), 15-28 (2004)
Isolation of trans, trans-2, 4-decadienal and intermedeol from cold-pressed citrus oils.
Moshonas MG & Shaw PE.
Journal of Agricultural and Food Chemistry, 27(1), 210-211 (1979)
Flavor volatiles in three rice cultivars with low levels of digestible protein during cooking.
Zeng Z, et al.
Cereal Chem., 85(5), 689-695 (2008)
Anna Lettieri et al.
Marine drugs, 13(3), 1451-1465 (2015-03-20)
The anti-proliferative effects of diatoms, described for the first time in copepods, have also been demonstrated in benthic invertebrates such as polychaetes, sea urchins and tunicates. In these organisms PUAs (polyunsaturated aldehydes) induce the disruption of gametogenesis, gamete functionality, fertilization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service