Skip to Content
Merck
All Photos(1)

Key Documents

W292508

Sigma-Aldrich

Propyl acetate

≥98%, FG

Synonym(s):

Acetic acid propyl ester, N-propyl ethanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH2CH3
CAS Number:
Molecular Weight:
102.13
FEMA Number:
2925
Beilstein:
1740764
EC Number:
Council of Europe no.:
192
MDL number:
UNSPSC Code:
12164502
eCl@ss:
39022103
PubChem Substance ID:
Flavis number:
9.002
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
biological source:
synthetic
food allergen:
no known allergens

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Halal
Kosher

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012
FDA 21 CFR 172.515

vapor density

3.5 (vs air)

vapor pressure

25 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

842 °F

expl. lim.

1.7 %, 37 °F
8 %

refractive index

n20/D 1.384 (lit.)

bp

102 °C (lit.)

mp

−95 °C (lit.)

density

0.888 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

celery; fruity; pear

SMILES string

CCCOC(C)=O

InChI

1S/C5H10O2/c1-3-4-7-5(2)6/h3-4H2,1-2H3

InChI key

YKYONYBAUNKHLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Thermodynamic Analysis of Eplerenone in 13 Pure Solvents at Temperatures from 283.15 to 323.15 K.: This study offers a detailed thermodynamic analysis of eplerenone in various solvents, including propyl acetate, highlighting its solubility characteristics and interactions at different temperatures (Liu et al., 2024).

  • Comparative Study on the H-Abstraction Reactions of Isopropyl Acetate and Propyl Acetate by HO(2) and OH Radicals.: This research compares the reactions of isopropyl acetate and propyl acetate with hydroxyl radicals, providing insights into their reactivity and potential applications in atmospheric chemistry (Gao et al., 2024).

  • Microemulsions of Nonionic Surfactant with Water and Various Homologous Esters: Preparation, Phase Transitions, Physical Property Measurements, and Application for Extraction of Tricyclic Antidepressant Drugs from Aqueous Media.: This study explores the use of microemulsions containing propyl acetate for the efficient extraction of drugs, demonstrating its application in enhancing the solubility and recovery rates of pharmaceutical compounds (Racovita et al., 2023).

Disclaimer

The substance W292508 has no REACH registration because the only supported use is the use in medicinal products for human or veterinary use or in food or feedingstuffs according to article 2 of the REACH Regulation (EC) No 1907/2006.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.2 °F - closed cup

Flash Point(C)

11.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nur-Bahiyah Abu-Bakar et al.
Journal of separation science, 39(5), 947-955 (2016-01-01)
A novel microextraction method based on vortex- and CO2 -assisted liquid-liquid microextraction with salt addition for the isolation of furanic compounds (5-hydroxymethyl-2-furaldehyde, 5-methyl-2-furaldehyde, 2-furaldehyde, 3-furaldehyde, 2-furoic and 3-furoic acids) was developed. Purging the sample with CO2 was applied after vortexing
Marwan Shalash et al.
Talanta, 174, 428-435 (2017-07-26)
A vortex-assisted liquid-liquid-liquid microextraction method followed by high performance liquid chromatography-diode array detection for the determination of fourteen phenolic acids (cinnamic, m-coumaric, chlorogenic, syringic, ferulic, o-coumaric, p-coumaric, vanillic, p-hydroxybenzoic, caffeic, 2, 4-dihydroxybenzoic, sinapic, gentisic and gallic acids) in honey, iced
A L Gustavsson et al.
Bioorganic & medicinal chemistry, 5(12), 2173-2183 (1998-02-12)
The enantiomers of cis- and trans-3-(4-propyl-cyclopent-2-enyl) propyl acetate, which are conformationally constrained analogues of (Z)-5-decenyl acetate (1), a sex pheromone component of the turnip moth, Agrotis segetum, have been synthesized and tested using the electrophysiological single-sensillum technique. The analogues mimic
Lan-Chun Nie et al.
Zhi wu sheng li yu fen zi sheng wu xue xue bao = Journal of plant physiology and molecular biology, 31(6), 663-667 (2005-12-20)
Aroma volatiles from apple (Malus domestica Borkh. var. Starkrimson) fruit at different stages of maturity were collected by solid adsorbent-Tenax-GC and determined by thermodesorption and GC-MS. Production of propyl acetate, butyl acetate, ethyl 2-methyl-butanoate and total ester volatiles and changes
[Gas chromatographic determination of propyl acetate in air].
N N Sirotkina et al.
Gigiena i sanitariia, (7)(7), 46-47 (1981-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service