Skip to Content
Merck
All Photos(1)

Key Documents

W290904

Sigma-Aldrich

Piperine

≥95%, FG

Synonym(s):

1-Piperoylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
FEMA Number:
2909
Beilstein:
90741
EC Number:
Council of Europe no.:
492
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
14.003
NACRES:
NA.21
grade:
FG
Halal
Kosher
biological source:
Piper nigrum L.
food allergen:
no known allergens

biological source

Piper nigrum L.

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥95%

form

powder, crystals or granules

mp

131-135 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

fatty; pepper

SMILES string

O=C(\C=C\C=C\c1ccc2OCOc2c1)N3CCCCC3

InChI

1S/C17H19NO3/c19-17(18-10-4-1-5-11-18)7-3-2-6-14-8-9-15-16(12-14)21-13-20-15/h2-3,6-9,12H,1,4-5,10-11,13H2/b6-2+,7-3+

InChI key

MXXWOMGUGJBKIW-YPCIICBESA-N

Gene Information

human ... CYP3A4(1576)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Piperine, a major component of black pepper extracts, shown to have potential for inhibiting fat cell formation.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janine Zaugg et al.
Journal of natural products, 73(2), 185-191 (2010-01-21)
A plant extract library was screened for GABA(A) receptor activity making use of a two-microelectrode voltage clamp assay on Xenopus laevis oocytes. An ethyl acetate extract of black pepper fruits [Piper nigrum L. (Piperaceae) 100 microg/mL] potentiated GABA-induced chloride currents
Ui-Hyun Park et al.
Journal of agricultural and food chemistry, 60(15), 3853-3860 (2012-04-03)
This study investigated the antiadipogenic activity of black pepper extract and its constituent piperine in 3T3-L1 preadipocytes as well as the underlying molecular mechanisms. Both black pepper extract and piperine, without affecting cytotoxicity, strongly inhibited the adipocyte differentiation of 3T3-L1
Hyejeong Jwa et al.
Biochemical pharmacology, 84(11), 1501-1510 (2012-09-25)
This study investigated the role of piperine in the transcriptional regulation of liver X receptor α (LXRα) and the effects of dietary piperine on high-fat diet (HFD)-induced hepatic steatosis and insulin resistance in mice. Furthermore, we explored the potential molecular
Paul B Yaffe et al.
Experimental and molecular pathology, 94(1), 109-114 (2012-10-16)
Piperine, an alkaloid phytochemical found in the fruit of black and long pepper plants, is reported to inhibit the growth of cancer cells; however, the mechanism of action in human cancer cells is not clear. In this study we investigated
Clara Grosso et al.
Phytochemical analysis : PCA, 24(2), 141-147 (2012-09-19)
Depression is a mental disease causing large personal and socio-economic problems, and new improved drugs are therefore needed. Selective monoamine oxidase A (MAO-A) inhibitors are potential anti-depressants, but discovering new MAO-A inhibitors from natural sources by bioassay-guided approaches are a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service